Skip to Content
Merck
All Photos(1)

Documents

310816

Sigma-Aldrich

D-Glucose-1-d1

98 atom % D

Synonym(s):

Deuterated Glucose, Labeled Glucose, Dextrose-1-d1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CHOH]4CDO
CAS Number:
Molecular Weight:
181.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

99% (CP)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

mp

150-152 °C (lit.)

mass shift

M+1

SMILES string

[2H]C1(O)O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i6D

InChI key

WQZGKKKJIJFFOK-QVTRYHEBSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomáš Pluskal et al.
Nature plants, 5(8), 867-878 (2019-07-25)
Kava (Piper methysticum) is an ethnomedicinal shrub native to the Polynesian islands with well-established anxiolytic and analgesic properties. Its main psychoactive principles, kavalactones, form a unique class of polyketides that interact with the human central nervous system through mechanisms distinct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service