Skip to Content
Merck
All Photos(1)

Key Documents

540277

Sigma-Aldrich

2,5-Dibromobenzotrifluoride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(Br)2CF3
CAS Number:
Molecular Weight:
303.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

mp

49-51 °C (lit.)

functional group

bromo
fluoro

SMILES string

FC(F)(F)c1cc(Br)ccc1Br

InChI

1S/C7H3Br2F3/c8-4-1-2-6(9)5(3-4)7(10,11)12/h1-3H

InChI key

VWKFJAOCLPPQGR-UHFFFAOYSA-N

General description

2,5-Dibromobenzotrifluoride is a halogenated hydrocarbon.

Application

2,5-Dibromobenzotrifluoride (1,4-Dibromo-2-(trifluoromethyl)benzene) may be used to synthesize 4-bromo-2-(trifluoromethyl)benzaldehyde and functionalized alkynyl-substituted (trifluoromethyl)benzenes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Site-Selective Sonogashira Reactions of 1, 4-Dibromo-2-(trifluoromethyl) benzene: Synthesis and Properties of Fluorinated Alkynylbenzenes.
Reimann S, et al.
European Journal of Organic Chemistry, 36, 8115-8134 (2013)
Highly efficient triarylene conjugated dyes for sensitized solar cells.
Chang YJ and Chow TJ.
Journal of Materials Chemistry, 21(26), 9523-9531 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service