Skip to Content
Merck
All Photos(2)

Documents

537632

Sigma-Aldrich

Isobutyl isobutyrate

≥98%

Synonym(s):

2-Methylpropyl 2-methylpropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
144.21
Beilstein:
1701355
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 39.9 °C)

Assay

≥98%

autoignition temp.

810 °F

refractive index

n20/D 1.398 (lit.)

bp

145-152 °C (lit.)

mp

−81 °C (lit.)

density

0.855 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC(=O)C(C)C

InChI

1S/C8H16O2/c1-6(2)5-10-8(9)7(3)4/h6-7H,5H2,1-4H3

InChI key

RXGUIWHIADMCFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boric acid catalyzed Tishchenko reactions.
Stapp PR.
The Journal of Organic Chemistry, 38(7), 1433-1434 (1973)
Response surface methodological approach for the synthesis of isobutyl isobutyrate.
Hamsaveni DR, et al.
Process. Biochem., 36(11), 1103-1109 (2001)
Yaoyao Peng et al.
Food research international (Ottawa, Ont.), 129, 108873-108873 (2020-02-11)
The phenolic compounds and aroma active compounds in feijoa (Acca sellowiana (O.Berg) Burret) juice from four New Zealand grown cultivars (Apollo, Unique, Opal Star, and Wiki Tu) were investigated. A high total phenolic content (maximum 1.89 mg GAE/mL juice) and significant
Béla P Molnár et al.
Frontiers in physiology, 9, 323-323 (2018-04-19)
Insects use sensitive olfactory systems to detect relevant host volatiles and avoid unsuitable hosts in a complex environmental odor landscape. Insects with short lifespans, such as gall midges (Diptera: Cecidomyiidae), are under strong selection pressure to detect and locate suitable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service