Skip to Content
Merck
All Photos(1)

Key Documents

58448

Sigma-Aldrich

2-Methyl-1-propanol

BioUltra, for molecular biology, ≥99.5% (GC)

Synonym(s):

Isobutanol, Isobutyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
1730878
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.26
grade:
Molecular Biology
for molecular biology
technique(s):
GC/GC: suitable
bp:
108 °C (lit.)
vapor pressure:
8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)

grade

Molecular Biology
for molecular biology

vapor density

2.55 (vs air)

vapor pressure

8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)

product line

BioUltra

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

801 °F

expl. lim.

10.6 %

technique(s)

GC/GC: suitable

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.001% peroxides (as H2O2)
≤0.005% free acid (as C3H7CHO)
≤0.01% aldehyde (as C3H7CHO)
≤0.01% ketone (as CH3COCH3)
≤0.1% water

evapn. residue

≤0.001%

refractive index

n20/D 1.396 (lit.)
n20/D 1.396

bp

108 °C (lit.)

mp

−108 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1.0 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CC(C)CO

λ

neat

UV absorption

λ: 260 nm Amax: 0.10
λ: 280 nm Amax: 0.06

InChI

1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

ZXEKIIBDNHEJCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Butanol concentration of DNA

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.M. Wallace
Methods in Enzymology, 152, 47-47 (1987)
R.C. Ogden et al.
Methods in Enzymology, 152, 68-68 (1987)
Antonino Baez et al.
Applied microbiology and biotechnology, 90(5), 1681-1690 (2011-05-07)
Promising approaches to produce higher alcohols, e.g., isobutanol, using Escherichia coli have been developed with successful results. Here, we translated the isobutanol process from shake flasks to a 1-L bioreactor in order to characterize three E. coli strains. With in
Jan-Karl Guterl et al.
ChemSusChem, 5(11), 2165-2172 (2012-10-23)
The limited supply of fossil resources demands the development of renewable alternatives to petroleum-based products. Here, biobased higher alcohols such as isobutanol are versatile platform molecules for the synthesis of chemical commodities and fuels. Currently, their fermentation-based production is limited
Aiqin Shi et al.
Metabolic engineering, 16, 1-10 (2012-12-19)
Isobutanol is an excellent alternative biofuel. Fermentative production of isobutanol had been realized in several microorganisms by combining branched-chain amino acids synthetic pathway and Ehrlich pathway. In contrast to using plasmid overexpression and inducible promoters, genetically stable Escherichia coli strains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service