Skip to Content
Merck
All Photos(1)

Documents

53310

Sigma-Aldrich

Histamine bisphosphate monohydrate

≥99.0% (NT)

Synonym(s):

2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(4-Imidazolyl)ethylamine diphosphate salt monohydrate, Histamine diphosphate salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3 · 2H3PO4 · H2O
CAS Number:
Molecular Weight:
325.15
Beilstein:
3744619
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (NT)

form

solid

mp

128-132 °C

storage temp.

−20°C

SMILES string

[H]O[H].OP(O)(O)=O.OP(O)(O)=O.NCCc1c[nH]cn1

InChI

1S/C5H9N3.2H3O4P.H2O/c6-2-1-5-3-7-4-8-5;2*1-5(2,3)4;/h3-4H,1-2,6H2,(H,7,8);2*(H3,1,2,3,4);1H2

InChI key

CNSGLKZGHLZDHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Histamine bisphosphate monohydrate is an imidazole containing heterocyclic building block that can be employed in chemical synthesis. Its crystal structure belongs to the monoclinic space group, P21/c and shows O–H ? O and N–H?O hydrogen bonds.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of histamine diphosphate monohydrate.
Veidis MV, et al.
J. Chem. Soc. Sect. A, 2659-2666 (1969)
L Shall et al.
Acta dermato-venereologica, 71(3), 199-204 (1991-01-01)
Terfenadine is a safe non-sedative H1-receptor antagonist. This study aimed to quantify the relative reduction in weal and flare area, thickness and erythema at 4, 8, 12 and 24 h following a single but variable oral dose of terfenadine compared
H S Nelson et al.
The Journal of allergy and clinical immunology, 92(5), 750-756 (1993-11-01)
Five commercially available devices for performing prick skin testing were compared for reproducibility, patient acceptance, occurrence of false-negative skin test results, and size distribution of reactions at the negative control sites. Reproducibility of skin testing with 10 mg/ml histamine base
L Shall et al.
Acta dermato-venereologica, 72(6), 451-453 (1992-11-01)
This study examined cutaneous mast cell behaviour in 14 patients with chronic urticaria but no dermographism and 11 healthy controls, by measuring cutaneous weal and flare reactions evoked in response to intradermal challenge injections of 0.1 ml isotonic saline, histamine
Y Larivée et al.
The Journal of otolaryngology, 30(6), 319-323 (2002-01-05)
The objectives of this study were to evaluate the reproducibility of the nasal response to histamine with the acoustic rhinometer and to compare the responses in normal and rhinitic subjects. Our study comprised 10 normal and 10 rhinitic subjects. Each

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service