Skip to Content
Merck
All Photos(2)

Key Documents

525170

Sigma-Aldrich

2,2,4-Trimethyl-1,3-pentanediol diisobutyrate

≥98.5%

Synonym(s):

1-Isopropyl-2,2-dimethyltrimethylene diisobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH[O2CCH(CH3)2]C(CH3)2CH2O2CCH(CH3)2
CAS Number:
Molecular Weight:
286.41
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.5%

autoignition temp.

795 °F

expl. lim.

0.48 %, 172 °F
3.1 %, 230 °F

refractive index

n20/D 1.434 (lit.)

bp

280 °C (lit.)

mp

−70 °C (lit.)

density

0.941 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CC(C)C(OC(=O)C(C)C)C(C)(C)COC(=O)C(C)C

InChI

1S/C16H30O4/c1-10(2)13(20-15(18)12(5)6)16(7,8)9-19-14(17)11(3)4/h10-13H,9H2,1-8H3

InChI key

OMVSWZDEEGIJJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TXIB is also referred to as Eastman plasticizer. 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate (TXIB) is widely used in food packaging. Its toxicity has been assessed.

Application

2,2,4-Trimethyl-1,3-pentanediol diisobutyrate may be used as a plasticizer in the fabrication of highly plasticized polyvinyl chloride, crosslinked by gamma irradiation.
Plasticizer.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

276.8 °F - Non-equilibrium method

Flash Point(C)

136 °C - Non-equilibrium method

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cutting out phthalates.
Tullo AH.
Chemical and Engineering News, 83(46) (2006)
Ulrike Bönisch et al.
PloS one, 7(7), e39817-e39817 (2012-07-18)
Epidemiological studies suggest an association between exposure to volatile organic compounds (VOCs) and adverse allergic and respiratory symptoms. However, whether VOCs exhibit a causal role as adjuvants in asthma development remains unclear. To investigate the effect of VOC exposure on
The toxicology and fate of 2,2,4-trimethyl-1,3-pentanediol diisobutyrate.
B D Astill et al.
Toxicology and applied pharmacology, 22(3), 387-399 (1972-07-01)
Radiation crosslinking of highly plasticized PVC.
Mendizabal E, et al.
Radiation Physics and Chemistry, 47(6), 305-309 (1996)
W S Cain et al.
Indoor air, 15(6), 445-457 (2005-11-05)
An experiment explored ability of subjects to detect vapors of the plasticizer TXIB (2,2,4-trimethyl-1,3-pentanediol diisobutyrate) and ethanol via olfaction and via ocular and nasal chemesthesis, i.e. chemically stimulated feel. Testing, tailored to the sensitivity of each subject, produced psychometric functions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service