Skip to Content
Merck
All Photos(1)

Documents

52360

Sigma-Aldrich

Hexadecyltrimethoxysilane

technical, ≥85% (GC)

Synonym(s):

Trimethoxyhexadecylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3C(CH2)15Si(OCH3)3
CAS Number:
Molecular Weight:
346.62
Beilstein:
1949851
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

technical

Assay

≥85% (GC)

form

liquid

impurities

isomers

refractive index

n20/D 1.44

density

0.89 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCC[Si](OC)(OC)OC

InChI

1S/C19H42O3Si/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23(20-2,21-3)22-4/h5-19H2,1-4H3

InChI key

RSKGMYDENCAJEN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hexadecyltrimethoxysilane (HDTMS) may be used to prepare hydrophobic silica aerogels by sol-gel process.{80} HDTMS was coated upon silica covered aluminum pigments.{81} Metal nanoparticles (CeO2, Fe2O4, TiO2, ZnO) coated with HDTMS form stable dispersions in hydrocarbon solvents. {82} HDTMS may be used to functionalize polyisocyanurate (PIR) foams {84} and graphene.{83}

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

247.1 °F - closed cup

Flash Point(C)

119.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corrosion protection of aluminum pigments by sol?gel coatings
Supplit R and Schubert U
Corrosion Science, 49(8), 3325-3332 (2007)
Organic modification of TEOS based silica aerogels using hexadecyltrimethoxysilane as a hydrophobic reagent
Nagaraja DH, et al.
Applied Surface Science, 253(3), 1566-1572 (2006)
Hexadecyltriethoxysilane-induced dispersions of metal oxide nanoparticles in nonpolar solvents.
Gomathi A and Rao CNR
Journal of Cluster Science, 19(1), 247-257 (2008)
Liquid nucleating additives for improving thermal insulating properties and mechanical strength of polyisocyanurate foams.
Kang MJ, et al.
J. Mater. Sci., 45(19), 5412-5419 (2010)
Preparation and characterization of long-chain alkyl silane-functionalized graphene film.
Ma WS, et al.
J. Mater. Sci., 48(1), 156-161 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service