Skip to Content
Merck
All Photos(2)

Key Documents

520705

Sigma-Aldrich

Platinum on alumina

greener alternative

extent of labeling: 0.5 wt. % loading, spheres

Synonym(s):

ESCAT 226, Pt/Al2O3

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

spheres

reaction suitability

core: platinum
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

0.5 wt. % loading

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Catalyst for:
  • In-situ ion exchange preparation of platinum-carbon nanotubes electrode
  • PtRu nanoparticles supported on nitrogen-doped porous carbon for use in fuel cells
  • Producing hydrogen from methylcyclohexane
  • Microfibrous structured catalytic packings for miniature methanol fuel processors

Used for:
  • Self-assembled monolayer of graphene-platinum counter electrode for efficient dye-sensitized solar cells
  • Borohydride interaction with metal surfaces
Platinum on alumina (Pt/Al2O3) (0.5 wt. % loading, spheres) can be used as a catalyst for a variety of applications, such as:
  • in the hydrodechlorination of chlorobenzenes
  • oxidation, reduction, and decomposition
  • in methanol steam reforming
  • hydrocarbon combustion

Legal Information

ESCAT is a trademark of BASF Catalysts, LLC

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. C. S. Escano, et al.,
The Journal of Physical Chemistry C, 115, 19883-19889 (2011)
F. Alhumaidan, et al.,
Energy and Fuels null
M. Ling, et al.,
International Journal of Hydrogen Energy, 36, 12833-12842 (2011)
Z. Liu, et al.,
ACS Applied Materials & Interfaces null
Feng Gong et al.
Physical chemistry chemical physics : PCCP, 13(39), 17676-17682 (2011-09-13)
Monolayer of PDDA/graphene/PDDA/H(2)PtCl(6) is fabricated on conductive glass using electrostatic layer-by-layer self-assembly technique, which is then converted to graphene/Pt monolayer for use as counter electrode in dye-sensitized solar cell (DSSC). As compared to the sputtered Pt counter electrode, the self-assembled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service