Skip to Content
Merck
All Photos(1)

Key Documents

517690

Sigma-Aldrich

1-Iodo-3-phenylpropane

97%

Synonym(s):

3-Iodopropylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)3I
CAS Number:
Molecular Weight:
246.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.5820 (lit.)

density

1.530 g/mL at 25 °C (lit.)

functional group

iodo
phenyl

SMILES string

ICCCc1ccccc1

InChI

1S/C9H11I/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2

InChI key

RGCKJSPKMTWLLX-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Zhang et al.
Journal of medicinal chemistry, 43(25), 4840-4849 (2000-12-22)
A series of analogues related to 1-[2-(diphenylmethoxy)ethyl]-4-(3-phenylpropyl)piperazine (2) and 1-¿2-[bis(4-fluorophenyl)methoxy]ethyl¿-4-(3-phenylpropyl)piperazine (3) (GBR 12935 and GBR 12909, respectively), in which the piperazine moiety was replaced by bridged piperazines for structural rigidity, has been designed, synthesized, and evaluated for their ability to
Thorsten Wilhelm et al.
Organic letters, 7(18), 4053-4056 (2005-08-27)
A new three-component, palladium-catalyzed domino reaction which gives access to meta-substituted arenes using aryl iodides and primary alkyl halides is reported. Various functional groups are tolerated on both the aryl iodide and alkyl halide. In addition, isotopic labeling studies provide
Synthesis of Aliphatic [carbonyl-11C] Esters Using [11C] Carbon Monoxide.
Itsenko O, et al.
European Journal of Organic Chemistry, 17, 3830-3834 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service