Skip to Content
Merck
All Photos(1)

Key Documents

140856

Sigma-Aldrich

3-Phenyl-1-propanol

98%

Synonym(s):

3-Phenylpropyl alcohol, Hydrocinnamyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)3OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
1857542
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

liquid

refractive index

n20/D 1.526 (lit.)

bp

119-121 °C/12 mmHg (lit.)

mp

−18 °C (lit.)

density

1.001 g/mL at 20 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

OCCCc1ccccc1

InChI

1S/C9H12O/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6,10H,4,7-8H2

InChI key

VAJVDSVGBWFCLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Phenyl-1-propanol is a fragrance ingredient[1].

Application

3-Phenyl-1-propanol was used to study the hydrogenation of trans-cinnamaldehyde using water-soluble organometallic complexes[2]. It was used as starting reagent during the enantioselective synthesis of (S)- and (R)-dapoxetine[3].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The regioselective biphasic hydrogenation of trans-cinnaldehyde by meta sulfonatophenyl-diphenylphosphine (TPPMS) Ru (II) and Os (II) species. The influence of ionic strength, ligand tensoactivity and metal nature in the selective production of the unsaturated alcohol.
Lopez-Linares F, et al.
J. Mol. Catal. A: Chem., 145(1), 61-68 (1999)
S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49 Suppl 2, S246-S251 (2011-08-23)
A toxicologic and dermatologic review of 3-phenyl-1-propanol when used as a fragrance ingredient is presented. 3-Phenyl-1-propanol is a member of the fragrance structural group cinnamyl phenylpropyl compounds. The common characteristic structural element of cinnamyl phenylpropyl materials is an aryl substituted
Jörgen Samuelsson et al.
Journal of chromatography. A, 1217(46), 7215-7221 (2010-10-12)
The elution by characteristic points (ECP) method is a very rapid and precise method for determination of the phase system equilibrium of phase systems in broad solute concentration ranges. Thus, the method is especially suitable for rapid characterization of high
Soyeong Kang et al.
The Journal of organic chemistry, 75(1), 237-240 (2009-12-05)
A highly efficient, enantioselective sequence has been developed for the synthesis of (S)- and (R)-dapoxetine. The pathways involve the intermediacy of the 6-membered-ring sulfamate esters 4, which were generated by Du Bois asymmetric C-H amination reactions of the prochiral sulfamate
E Baciocchi et al.
European journal of biochemistry, 268(3), 665-672 (2001-02-13)
The H(2)O(2)-promoted oxidations of (R)-[alpha-(2)H(1)]-and (S)-[alpha-(2)H(1)]-arylalkanols catalysed by chloroperoxidase (CPO) from Caldariomyces fumago have been investigated. It has been found that with (R)-[alpha-(2)H(1)]-alcohols, the oxidation involves almost exclusively the cleavage of the C-H bond, whereas in the case of the

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service