512990
2,4,6,8-Tetramethylcyclotetrasiloxane
≥98.5%, ≥99.999% trace metals basis
Synonym(s):
1,3,5,7-Cyclotetra(methylsiloxane), 1,3,5,7-Tetramethylcyclotetrasiloxane, TMCTS
About This Item
Recommended Products
Assay
≥98.5%
≥99.999% trace metals basis
form
liquid
refractive index
n20/D 1.387 (lit.)
bp
134 °C (lit.)
mp
−69 °C (lit.)
density
0.986 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
C[SiH]1O[SiH](C)O[SiH](C)O[SiH](C)O1
InChI
1S/C4H16O4Si4/c1-9-5-10(2)7-12(4)8-11(3)6-9/h9-12H,1-4H3
InChI key
BQYPERTZJDZBIR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
75.2 °F - closed cup
Flash Point(C)
24 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service