Skip to Content
Merck
All Photos(2)

Key Documents

494070

Sigma-Aldrich

6,6′′-Dibromo-2,2′:6′,2′′-terpyridine

90%

Synonym(s):

6,6′′-Dibromo-2,2′:6′,2′′-terpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9Br2N3
CAS Number:
Molecular Weight:
391.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

90%

mp

257-263 °C (lit.)

functional group

bromo

SMILES string

Brc1cccc(n1)-c2cccc(n2)-c3cccc(Br)n3

InChI

1S/C15H9Br2N3/c16-14-8-2-6-12(19-14)10-4-1-5-11(18-10)13-7-3-9-15(17)20-13/h1-9H

InChI key

PYMBATDYUCQLBC-UHFFFAOYSA-N

General description

6,6′′-Dibromo-2,2′:6′,2′′-terpyridine is a substituted terpyridine that can be synthesized from 2,6-dibromopyridine.[1]

Application

6,6′′-Dibromo-2,2′:6′,2′′-terpyridine may be used in the synthesis of a novel disubstituted terpyridine ligand.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Novel One-Pot Synthesis of 6,6''-Dibromo-2,2':6'2''-terpyridine.
Uchida Y, et al.
ChemInform, 26(50) (1995)
Lingjing Chen et al.
Dalton transactions (Cambridge, England : 2003), 48(26), 9596-9602 (2019-02-20)
A new pentadentate quinoline-pyridine ligand and its iron (1), cobalt (2) and nickel (3) complexes have been synthesized and characterized. The iron complex exhibits excellent photocatalytic activity towards CO2-to-CO conversion with a TON(CO) of 544 and a selectivity of 99.3%
Kogularamanan Suntharalingam et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(47), 15133-15141 (2012-10-13)
A new disubstituted terpyridine ligand and the corresponding tri-copper(II) complex have been prepared and characterised. The binding affinity and binding mode of this tri-copper complex (as well as the previously reported mono- and di-copper analogues) towards duplex DNA were determined

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service