481874
3-tert-Butyl-2-hydroxybenzaldehyde
96%
Synonym(s):
3-tert-Butylsalicylaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)3CC6H3(OH)CHO
CAS Number:
Molecular Weight:
178.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
96%
refractive index
n20/D 1.544 (lit.)
bp
78-79 °C/1 mmHg (lit.)
density
1.041 g/mL at 25 °C (lit.)
functional group
aldehyde
SMILES string
CC(C)(C)c1cccc(C=O)c1O
InChI
1S/C11H14O2/c1-11(2,3)9-6-4-5-8(7-12)10(9)13/h4-7,13H,1-3H3
InChI key
ROILLNJICXGZQQ-UHFFFAOYSA-N
Related Categories
General description
3-tert-Butyl-2-hydroxybenzaldehyde is a mono-tert-butyl substituted 2-hydroxybenzaldehyde that can be synthesized using ethylbromide and 2-tert-butylphenol.
Application
3-tert-Butyl-2-hydroxybenzaldehyde may be used in the synthesis of:
- 5-bromo-3-tert-butyl-2-hydroxybenzaldehyde
- 3-tert-butyl-5-(chloro-methyl)-2- hydroxybenzaldehyde
- chiral Schiff base-titanium alkoxide catalysts
- 2 -(3-tert-butyl -2-hydroxybenzyl)aminomethylpyridine
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Karuthamohamed Dhahagani et al.
Materials science & engineering. C, Materials for biological applications, 90, 119-130 (2018-06-02)
New morpholine derived Schiff base ligands (HL1 and HL2) and their Cu(II) complexes [Cu(L1)2] (1) and [Cu(L2)2] (2) have been synthesized and characterized by 1H NMR, IR, UV-Vis, EPR studies and cyclic voltammetric analyses. Single crystal X-ray crystallography studies confirm
Enantioselective trimethylsilylcyanation of some aldehydes catalyzed by chiral Schiff base-titanium alkoxide complexes.
Hayashi M, et al.
The Journal of Organic Chemistry, 58(6), 1515-1522 (1993)
Catalytic Asymmetric Cycloaddition of Carbon Dioxide and Propylene Oxide Using Novel Chiral Polymers of BINOL-Salen-Cobalt (III) Salts.
Yan P and Jing H.
Advanced Synthesis & Catalysis, 351(90, 1325-1332 (2009)
A novel polymeric chiral salen Mn (III) complex as solvent-regulated phase transfer catalyst in the asymmetric epoxidation of styrene.
Tan R, et al.
J. Mol. Catal. A: Chem., 259(1), 125-132 (2006)
New dioxo-molybdenum (vi) and-tungsten (vi) complexes with N-capped tripodal N2O2 tetradentate ligands: Synthesis, structures and catalytic activities towards olefin epoxidation.
Wong YL, et al.
Dalton Transactions, 39(19), 4602-4611 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service