Skip to Content
Merck
All Photos(2)

Documents

481556

Sigma-Aldrich

Iodine monochloride

99.998% trace metals basis

Synonym(s):

Chloroiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICl
CAS Number:
Molecular Weight:
162.36
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.998% trace metals basis

impurities

<25 ppm total metallic impurities

bp

97.4 °C (lit.)

density

3.24 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClI

InChI

1S/ClI/c1-2

InChI key

QZRGKCOWNLSUDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Meets A.C.S. reagent specifications.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Khalil et al.
Nucleosides, nucleotides & nucleic acids, 33(12), 786-799 (2014-11-06)
The reaction of thymidine, 3-mono-, and 3,3',5'-trialkylsubstitued thymidine analogues with iodine monochloride (ICl) was investigated. Treatment with ICl resulted in rapid deglycosylation, anomerization, and isomerization of thymidine and 3-substituted thymidine analogues under various reaction conditions leading to the formation of
Cation radicals as intermediates in aromatic halogenation with iodine monochloride: solvent and salt effects on the competition between chlorination and iodination.
Hubig SM, et al.
The Journal of Organic Chemistry, 59(21), 6233-6244 (1994)
A B Mason et al.
The Biochemical journal, 247(2), 417-425 (1987-10-15)
Iodination of the C-terminal half-molecule domain of ovotransferrin (OTF) causes a significant reduction in binding to transferrin receptors on chick reticulocytes when compared to the binding observed with holo-OTF or the N-terminal half-molecule domain. (In such studies binding of iodinated
R G Nielsen et al.
Journal of chromatography, 423, 41-50 (1987-12-25)
Low-level adsorption on the stationary phase has been studied using immunochemical reagents. An immunoaffinity column has been evaluated using affinity-purified radioisotope-labeled monoclonal antibodies. Recovery experiments including continuous immunosorbent monitoring have been performed. Proper characterization of an immunoaffinity separation can result
Ru-Jin Huang et al.
Analytical chemistry, 81(5), 1777-1783 (2009-02-10)
This study concerns the development of a coupled diffusion denuder system capable of separating and quantifying gaseous molecular iodine (I(2)) and two other highly reactive iodine species, ICl and HOI, which are collectively named activated iodine compounds (AIC). Both I(2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service