Skip to Content
Merck
All Photos(1)

Key Documents

47624

Sigma-Aldrich

Fmoc-Lys(Boc)-OH

≥98.0% (HPLC), for peptide synthesis

Synonym(s):

Nα-Fmoc-Nε-Boc-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32N2O6
CAS Number:
Molecular Weight:
468.54
Beilstein:
4217767
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

Fmoc-Lys(Boc)-OH, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

form

solid

optical activity

[α]20/D −12±1°, c = 1% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C26H32N2O6/c1-26(2,3)34-24(31)27-15-9-8-14-22(23(29)30)28-25(32)33-16-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h4-7,10-13,21-22H,8-9,14-16H2,1-3H3,(H,27,31)(H,28,32)(H,29,30)/t22-/m0/s1

InChI key

UMRUUWFGLGNQLI-QFIPXVFZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fmoc-Lys(Boc)-OH is an amino acid derivative used as a reagent in Fmoc solid-phase peptide synthesis.

Application

Fmoc-Lys(Boc)-OH can be used for:
  • The solid-phase synthesis of pentasubstituted dihydroimidazolylbutyl dihydroimidazol-3-ium salts by coupling with p-methylbenzhydrylamine (MBHA) resin.
  • Fmoc-based peptide synthesis of bis-naphthalene diimide, a threading intercalator.
  • Synthesis of ε-Boc-ε-(3,5-bis-trifluoromethyl-benzyl)-α-Fmoc-L-Lysine to be used as 19F NMR based screening tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-phase parallel synthesis of substituted dihydroimidazolylbutyl dihydroimidazol-3-ium salts.
Acharya AN, et al.
Tetrahedron Letters, 43(7), 1157-1160 (2002)
Improved synthesis of polyfluorinated l-lysine for 19F NMR-based screening.
Malgesini B, et al.
Molecular Diversity, 13(1), 53-56 (2009)
Marta Kulik et al.
Biochimie, 138, 32-42 (2017-04-12)
A fragment of 23S ribosomal RNA (nucleotides 1906-1924 in E. coli), termed Helix 69, forms a hairpin that is essential for ribosome function. Helix 69 forms a conformationally flexible inter-subunit connection with helix 44 of 16S ribosomal RNA, and the nucleotide
Marine Bacchi et al.
Protein science : a publication of the Protein Society, 25(12), 2225-2242 (2016-09-28)
Synthetic biology (or chemical biology) is a growing field to which the chemical synthesis of proteins, particularly enzymes, makes a fundamental contribution. However, the chemical synthesis of catalytically active proteins (enzymes) remains poorly documented because it is difficult to obtain
Bis-naphthalene diimide exhibiting an effective bis-threading intercalating ability.
Nojima T, et al.
Nucleic Acids Symposium Series, 3(1), 123-124 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service