Skip to Content
Merck
All Photos(2)

Key Documents

474576

Sigma-Aldrich

Poly(4-vinylphenol-co-methyl methacrylate)

greener alternative

Synonym(s):

PVP-co-PMMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H4OH)]x[CH2C(CH3)(CO2CH3)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

monomer molar fraction = ~ 50%

form

solid

mol wt

Mn 3,000-5,000 Da
average Mw 8.000-12,000 Da

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

224 °C (lit.)

density

1.3 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

COC(=O)C(C)=C.Oc1ccc(C=C)cc1

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Application

Used in electronics for solder resists, etching resists, presensitized printing plates, coatings and adhesives.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

581.0 °F - closed cup

Flash Point(C)

305 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service