Skip to Content
Merck
All Photos(2)

Key Documents

471747

Sigma-Aldrich

tert-Butyl N-(2,3-dihydroxypropyl)carbamate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2NHCO2C(CH3)3
CAS Number:
Molecular Weight:
191.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

60-63 °C (lit.)

SMILES string

CC(C)(C)OC(=O)NCC(O)CO

InChI

1S/C8H17NO4/c1-8(2,3)13-7(12)9-4-6(11)5-10/h6,10-11H,4-5H2,1-3H3,(H,9,12)

InChI key

OWAMQHJPVUGZSB-UHFFFAOYSA-N

General description

tert-Butyl N-(2,3-dihydroxypropyl)carbamate is a protected amine.

Application

tert-Butyl N-(2,3-dihydroxypropyl)carbamate may be used in the preparation of isobaric mix solution for use in post-column infusion experiment during single-stage orbitrap mass spectrometric analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kaufmann et al.
Rapid communications in mass spectrometry : RCM, 26(9), 1081-1090 (2012-04-03)
Orbitrap technology offers a combination of different technical specifications which have not yet been achieved by other high-resolution mass spectrometry instrumentation. This refers to the combination of sensitivity, dynamic range, mass accuracy, resolution and speed. The high stability of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service