Skip to Content
Merck
All Photos(2)

Key Documents

471453

Sigma-Aldrich

2-Dimethylaminoethanol

≥99.5%

Synonym(s):

N,N-Dimethyl-2-hydroxyethylamine, N,N-Dimethylethanolamine, Jeffcat DMEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
1209235
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

100 mmHg ( 55 °C)
6.12 mmHg ( 20 °C)

Assay

≥99.5%

refractive index

n20/D 1.4294 (lit.)

bp

134-136 °C (lit.)

mp

−70 °C (lit.)

solubility

H2O: soluble

density

0.886 g/mL at 20 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CN(C)CCO

InChI

1S/C4H11NO/c1-5(2)3-4-6/h6H,3-4H2,1-2H3

InChI key

UEEJHVSXFDXPFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Dimethylaminoethanol (deanol, DMAE) is a precursor of acetylcholine.[1][2] Microwave spectral studies on DMAE have reported the following values; the rotational constants (MHz) A = 5814.0(2), B = 2214.54(2), and C = 2037.96(2) and a dipole moment of 2.56 D, with a, b, and c components (D) of 2.27(2), 0.3(1), and 1.16(5), respectively.[3]

Application

2-Dimethylaminoethanol (deanol, DMAE) may be employed as a ligand in the copper-catalyzed amination of aryl bromides and iodides.[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - Seta closed cup

Flash Point(C)

40 °C - Seta closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occurrence and quantitative determination of 2-dimethylaminoethanol in animal tissue extracts.
C G HONEGGER et al.
Nature, 184(Suppl 8), 550-552 (1959-08-15)
Split face study on the cutaneous tensile effect of 2-dimethylaminoethanol (deanol) gel.
Uhoda I, et al.
Skin Research and Technology, 8(3), 164-167 (2002)
Copper-catalyzed aryl amination in aqueous media with 2-dimethylaminoethanol ligand.
Lu Z and Twieg RJ.
Tetrahedron Letters, 46(17), 2997-3001 (2005)
Microwave spectrum and hydrogen bonding in 2-dimethylaminoethanol.
Penn RE and Birkenmeier JA
Journal of Molecular Spectroscopy, 62(3), 416-422 (1976)
B Sommerfeld
Phytomedicine : international journal of phytotherapy and phytopharmacology, 14(11), 711-715 (2007-10-10)
Tricutan is a combination product of herbal extracts traditionally used for treatment of skin conditions, together with dimethylaminoethanol. The effectiveness of Tricutan in improving skin firmness and elasticity in photoaged facial skin was examined in a randomised, placebo-controlled, double-blind, split-face

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service