All Photos(2)
About This Item
Linear Formula:
CH3COC6H4B(OH)2
CAS Number:
Molecular Weight:
163.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95%
impurities
<10% 3-acetylphenylboronic anhydride
mp
204-208 °C (lit.)
functional group
ketone
SMILES string
CC(=O)c1cccc(c1)B(O)O
InChI
1S/C8H9BO3/c1-6(10)7-3-2-4-8(5-7)9(11)12/h2-5,11-12H,1H3
InChI key
SJGGDZCTGBKBCK-UHFFFAOYSA-N
Application
3-Acetylphenylboronic acid can be used as a substrate:
- In the synthesis of symmetric biaryls via oxidative dimerization using a palladium catalyst and water as a solvent.
- In the synthesis of aryl fluorides through electrophilic fluorination reaction using acetyl hypofluorite.
- In the coupling reactions of organoboranes with olefins using molecular oxygen and palladium catalyst.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Fluorination of aryl boronic acids using acetyl hypofluorite made directly from diluted fluorine
Vints I, et al.
The Journal of Organic Chemistry, 78(23), 11794-11797 (2013)
Oxygen-promoted Pd (II) catalysis for the coupling of organoboron compounds and olefins
Jung YC, et al.
Organic Letters, 5(13), 2231-2234 (2003)
Oxidative dimerization: Pd (II) catalysis in the presence of oxygen using aqueous media
Parrish JP, et al.
Tetrahedron Letters, 43(44), 7899-7902 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service