Skip to Content
Merck
All Photos(1)

Documents

469599

Sigma-Aldrich

Ethyl chlorodifluoroacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCF2CO2C2H5
CAS Number:
Molecular Weight:
158.53
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.358 (lit.)

bp

96-97.5 °C (lit.)

density

1.252 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(F)(F)Cl

InChI

1S/C4H5ClF2O2/c1-2-9-3(8)4(5,6)7/h2H2,1H3

InChI key

GVCAWQUJCHZRCB-UHFFFAOYSA-N

General description

Ethyl chlorodifluoroacetate (ECDFA) undergoes Reformatskii reaction with various aldehydes in DMF. It reacts with phenylacetylene to afford ethyl α,α−difluoro-4-phenyl-3-butenoates.

Application

Ethyl chlorodifluoroacetate (ECDFA) may be used as a starting reagent in the synthesis of 3-gem-difluoro-2-ethoxy allylic alcohols.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A theoretical investigation on the kinetics and reactivity of the gas-phase reactions of ethyl chlorodifluoroacetate with OH radical and Cl atom at 298 K.
Mishra BK, et al.
Structural Chemistry, 25(2), 463-470 (2014)
Fluorine-containing organozinc reagents. IV.: Reformatskii-type reactions of chlorodifluoroacet1c acid derivatives.
Lang RW and Schaub B.
Tetrahedron Letters, 29(24), 2943-2946 (1988)
Synthesis of 3-gem-difluoro-2-ethoxy allylic alcohols from ethyl chlorodifluoroacetate.
Begue J-P, et al.
Tetrahedron Letters, 35(3), 6097-6100 (1994)
Wadih Ghattas et al.
The Journal of organic chemistry, 71(22), 8618-8621 (2006-10-27)
An efficient preparation of pure ethyl Z- and E-alpha,alpha-difluoro-4-phenyl-3-butenoate 1a and 1b together with the corresponding acids 2a and 2b is described. The procedures involve stereocontrolled additions of *CF2CO2Et to phenylacetylene or beta-bromostyrene. Compound 1a is easily obtained by addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service