Recommended Products
Assay
98%
mp
154-157 °C (lit.)
SMILES string
OC(=O)COc1ccc(O)cc1
InChI
1S/C8H8O4/c9-6-1-3-7(4-2-6)12-5-8(10)11/h1-4,9H,5H2,(H,10,11)
InChI key
PKGWLCZTTHWKIZ-UHFFFAOYSA-N
General description
(4-Hydroxyphenoxy)acetic acid along with formaldehyde ammonium salt forms a polyaromatic anionic compound RG-13577. RG-13577 mimics the synthetic heparin and specifically binds to the vascular smooth muscle cells (SMCs) and inhibits their proliferative growth. In the molecules of (4-hydroxyphenoxy)acetic acid, the carboxyl groups are held together by R22(8) hydrogen bonds.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
(4-Hydroxyphenoxy) acetic acid.
Acta Crystallographica Section E, Structure Reports Online, 63(6), o2931-o2931 (2007)
Journal of cellular biochemistry, 81(1), 114-127 (2001-02-17)
A synthetic heparin-mimicking polyaromatic anionic compound RG-13577 (polymer of 4-hydroxyphenoxy acetic acid and formaldehyde ammonium salt, Mr approximately 5800) exhibits specific binding to vascular smooth muscle cells (SMCs) and inhibits their proliferative response to growth promoting factors. Receptor binding of
Bioorganic & medicinal chemistry letters, 12(13), 1709-1713 (2002-06-18)
An epoxybenzoquinone, 4-hydroxyphenoxypropionic acid, and 2-hydroxy-3-phenyl-3-butenoic acid derivatives have been designed, synthesized, and evaluated for in vitro inhibition activity against 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) from pig liver by the spectrophotometric enol-borate method. The biological data demonstrated that neither epoxybenzoquinone ester nor
Analytical biochemistry, 179(2), 288-290 (1989-06-01)
A fast and sensitive spectrophotometric method has been developed to measure the level of hydroxylated by-products (p-hydroxyphenoxyacetic acid and p-hydroxypenicillin V) in penicillin V fermentations. The method is based on a color reaction of the above-mentioned phenolic by-products with nitrous
Colloids and surfaces. B, Biointerfaces, 102, 95-101 (2012-09-26)
Individual dispersion of single-walled carbon nanotubes (SWNTs) in biocompatible media is of particular interest for diverse biomedical and nanomedicine applications. Herein we present, for the first time, a neutral pH water-soluble chitosan derivative, chitosan-hydroxyphenyl acetamide (CHPA), prepared by functionalizing the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service