Skip to Content
Merck
All Photos(2)

Key Documents

467863

Sigma-Aldrich

Lead(II) acetate trihydrate

≥99.99% trace metals basis

Synonym(s):

Lead diacetate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)2 · 3H2O
CAS Number:
Molecular Weight:
379.33
Beilstein:
3730298
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.99% trace metals basis

form

solid

reaction suitability

core: lead
reagent type: catalyst

impurities

≤0.01% insolubles
<100 ppm total metallic impurities

mp

75 °C (dec.) (lit.)

anion traces

NO3- and NO2-: ≤0.005%
chloride (Cl-): ≤5 ppm

cation traces

Ca: ≤0.005%
Cu: ≤0.002%
Fe: ≤0.001%
K: ≤0.005%
Na: ≤0.01%

SMILES string

O.O.O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2

InChI key

MCEUZMYFCCOOQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Lead(II) acetate trihydrate is a widely used in the field of textile dyeing, varnishes, waterproofing and insecticides.

Application

Desulfurization reagent; also used as neutralization of HCl.
Lead(II) acetate trihydrate is used:
  • In the preparation of lead lanthanum zirconate stannate titanate (PbLa(ZrSnTi)O3) antiferroelectric (AFE) ceramics for dielectric capacitors
  • As a precursor to synthesize lead chalcogenides such as lead sulfide (PbS), lead selenide (PbSe) and lead telluride (PbTe) nanostructures.
  • In the synthesis of organic-inorganic halide perovskites (OIHPs) as optoelectronic materials for solar cells and light-emitting diodes.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple cubic super crystals containing PbTe nanocubes and their core- shell building blocks.
Zhang J, et al.
Journal of the American Chemical Society, 130(45), 15203-15209 (2008)
High-energy density of Pb0. 97La0. 02 (Zr0. 50Sn0. 45Ti0. 05) O3 antiferroelectric ceramics prepared by sol-gel method with low-cost dibutyltin oxide.
Dan Y, et al.
Journal of the American Ceramic Society. American Ceramic Society, 0(0) (2018)
Infiltration of methylammonium metal halide in highly porous membranes using sol-gel-derived coating method.
Kwon SL, et al.
Applied Surface Science, 416(45), 96-102 (2017)
Designing PbSe nanowires and nanorings through oriented attachment of nanoparticles.
Cho K-S, et al.
Journal of the American Chemical Society, 127(19), 7140-7147 (2005)
Cragg, R. H.; Weston, A. F.
Tetrahedron Letters, 655-655 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service