Skip to Content
Merck
All Photos(1)

Key Documents

462861

Sigma-Aldrich

Iron(III) p-toluenesulfonate hexahydrate

technical grade

Synonym(s):

Iron (III) 4-methyl-benzenesulfonic acid salt hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4SO3)3Fe · 6H2O
CAS Number:
Molecular Weight:
677.52
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

reaction suitability

core: iron
reagent type: catalyst

SMILES string

O.O.O.O.O.O.[Fe+3].Cc1ccc(cc1)S([O-])(=O)=O.Cc2ccc(cc2)S([O-])(=O)=O.Cc3ccc(cc3)S([O-])(=O)=O

InChI

1S/3C7H8O3S.Fe.6H2O/c3*1-6-2-4-7(5-3-6)11(8,9)10;;;;;;;/h3*2-5H,1H3,(H,8,9,10);;6*1H2/q;;;+3;;;;;;/p-3

InChI key

FACZRMXNSYHQMG-UHFFFAOYSA-K

Application

Iron(III) p-toluenesulfonate hexahydrate (Fe(CH3C6H4SO3)3·6H2O) can be used:
  • As an oxidant in the synthesis of poly(3,4-ethylenedioxythiophene) (PEDOT) nanofilms by vapor phase polymerization.
  • In the synthesis of iron(III) complex of pyridoxal-4-methylthiosemicarbazone.
  • As a precursor material for the synthesis of Fe, N, and S functionalized carbon electrocatalyst for the oxygen reduction reaction.
  • As an oxidant in the synthesis of poly(thiophene phenylenes) (PThP).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effectively incorporating iron, nitrogen, and sulfur functionalities on carbon surface for a superior electrocatalyst toward oxygen reduction reaction
Zhang W, et al.
Electrochemical Communications, 81(13), 34-37 (2017)
Effects of iron (III) p-toluenesulfonate hexahydrate oxidant on the growth of conductive poly (3, 4-ethylenedioxythiophene)(PEDOT) nanofilms by vapor phase polymerization
Ali MA, et al.
Synthetic Metals, 161(13-14), 1347-1352 (2011)
Highly functionalisable polythiophene phenylenes
Chan EWC, et al.
Polym. Chem., 6(43), 7618-7629 (2015)
Crystal structure and magnetic behaviour of a five-coordinate iron (III) complex of pyridoxal-4-methylthiosemicarbazone
Tido EWY, et al.
Inorgorganica Chimica Acta, 360(13), 3896-3902 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service