Skip to Content
Merck
All Photos(1)

Key Documents

457647

Sigma-Aldrich

Poly(D,L-lactide-co-caprolactone)

DL-lactide 86 mol %

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

feed ratio

lactide:caprolactone 85:15

composition

DL-lactide, 86 mol %

inherent viscosity

0.7-0.9 dL/g, in chloroform(lit.)

transition temp

Tg 16 °C

storage temp.

2-8°C

SMILES string

CC(O)C(O)=O.OCCCCC(O)=O

InChI

1S/C5H10O3.C3H6O3/c6-4-2-1-3-5(7)8;1-2(4)3(5)6/h6H,1-4H2,(H,7,8);2,4H,1H3,(H,5,6)

InChI key

LLHKFIFJPCHEOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(DL-lactide-co-caprolactone) (PLA-PCL) is a biodegradable block copolymer that may be used as monofilament sutures and nanoparticle-based drug delivery systems.

Features and Benefits

Biocompatible, biodegradable polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymeric scaffolds in tissue engineering application: a review.
Dhandayuthapani B, et al.
International Journal of Polymer Science (2011)
Novel docetaxel-loaded nanoparticles based on poly (lactide-co-caprolactone) and poly (lactide-co-glycolide-co-caprolactone) for prostate cancer treatment: formulation, characterization, and cytotoxicity studies.
Sanna V, et al.
Nanoscale Research Letters, 6(1), 260-260 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service