Skip to Content
Merck
All Photos(1)

Key Documents

568562

Sigma-Aldrich

Poly(L-lactide-co-caprolactone-co-glycolide)

L-lactide 70 %, average Mn ~50,000 by GPC

Synonym(s):

ε-Caprolactone-glycolide-L -lactide copolymer, Poly-L -lactide-co-glycolide-co-ε-caprolactone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

feed ratio

lactide:caprolactone:glycolide 70:20:10

mol wt

average Mn ~50,000 by GPC

composition

L-lactide, 70%

transition temp

Tg ~28 °C

storage temp.

2-8°C

Application

Poly(L-lactide-co-caprolactone-co-glycolide) (PLGA-PCL) is a biodegradable block copolymer that may be used as nanoparticle-based drug delivery systems. Electrospun nanofiber blend of PLGA-PCL, phosphorylcholine groups and an antiproliferative (rapamycin)may be used as vascular grafts and prostheses.

Features and Benefits

Biocompatible, biodegradable polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of electrospun poly (L-lactide-co-caprolactone-co-glycolide)/phospholipid polymer/rapamycin blended fibers for vascular application.
Kim H I, et al.
Current Applied Physics, 9(4), e249-e251 (2009)
Novel docetaxel-loaded nanoparticles based on poly (lactide-co-caprolactone) and poly (lactide-co-glycolide-co-caprolactone) for prostate cancer treatment: formulation, characterization, and cytotoxicity studies.
Sanna V, et al.
Nanoscale Research Letters, 6(1), 260-260 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service