Skip to Content
Merck
All Photos(3)

Key Documents

452890

Sigma-Aldrich

Sodium borohydride

caplets (18 × 10 × 8 mm), 98%

Synonym(s):

Vertellus, Sodium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

caplets (18 × 10 × 8 mm)

contains

≤100 ppm MgCO3 as anticaking agent

reaction suitability

reagent type: reductant

mp

>300 °C (dec.) (lit.)

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium borohydride is a mild reducing agent commonly used for the reduction of aldehydes and ketones to their corresponding alcohols.[1][2] It is also a promising candidate as a hydrogen carrier and storage media for polymer electrolyte membrane (PEM) fuel cells.[3][4] NaBH4 caplets can be used for the synthesis of Ti3+-doped titanium dioxide powders from TiO2 powders.[5]

Legal Information

Vertellus product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sodium borohydride
e-EROS Encyclopedia of Reagents for Organic Synthesis., 1-13 (2014)
Controllable synthesis and tunable photocatalytic properties of Ti3+-doped TiO2
Ren, Ren and Wen, Zhenhai and Cui, Shumao and Hou, Yang and Guo, Xiaoru and Chen, Junhong
Scientific Reports, 5, 10714-10714 (2015)
Evaluation of Acid Accelerators for an Off-Grid PEM Fuel Cell Power Station Utilizing Solid Sodium Borohydride for Hydrogen Storage
Hsu L, et al.
ECS Transactions, 75(14), 515-520 (2016)
Hydrogen production by hydrolysis of sodium borohydride for PEM fuel cells feeding
International Workshop, Advances in Fuel Cells and the Hydrogen Economy (2008)
Forty years of hydride reductions.
Brown H C and Krishnamurthy S
Tetrahedron, 35(5), 567-607 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service