Skip to Content
Merck
All Photos(3)

Key Documents

480886

Sigma-Aldrich

Sodium borohydride

greener alternative

granular, 99.99% trace metals basis

Synonym(s):

Sodium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99.99% trace metals basis

form

granular

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>300 °C (dec.) (lit.)

greener alternative category

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Nanocrystalline superlattices in gold colloid solution have been prepared by ligand-induction using AuCl3 reduced with sodium borohydride.[1] Nucleophilic addition of hydride ion from sodium borohydride is an inexpensive alternative method for the Baylis-Hillman reaction to form [E]-a-methylcinnamic acids.[2] A reductant solution composed of NaBH4 and NaOH was used to form mercury cold vapor.[1] NaBH4 may be used to generate hydrogen by catalytic hydrolysis.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Mercury in Fly Ash by Using Flow Injection Cold Vapor Isotope Dilution Inductively Coupled Plasma Mass Spectrometry.
Suh, Jung-Ki, et al.
Mass Spectrom., 3(2), 58-61 (2012)
An effective nanosensor for organic molecules based on water-soluble mercaptopropionic acid-capped CdTe nanocrystals with potential application in high-throughput screening and high-resolution optical microscopy
Lau, Pick-Chung, et al.
Biomedical Optics Express, 5(7), 2420-2434 (2014)
Sandwich Enzyme--linked Immunosorbent Assay (ELISA) Analysis of Plant Cell Wall Glycan Connections.
Cornuault V and Knox JP
Bio-protocol, 4(8) (2014)
Basavaiah, D. et al.
The Journal of Organic Chemistry, 64, 1197-1197 (1999)
Metal Borohydrides as Hydrogen Storage Materials
Soloveichik GL
Material Matters, 2(2), 11-11 (2007)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service