Skip to Content
Merck
All Photos(1)

Documents

451398

Sigma-Aldrich

Zinc bromide

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Zinc dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnBr2
CAS Number:
Molecular Weight:
225.20
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38150410
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.999% trace metals basis

reaction suitability

reagent type: catalyst
core: zinc

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

bp

~670 °C/1 atm (lit.)

mp

394 °C (lit.)

solubility

diethyl ether: slightly soluble(lit.)
ethanol: very soluble(lit.)

SMILES string

Br[Zn]Br

InChI

1S/2BrH.Zn/h2*1H;/q;;+2/p-2

InChI key

VNDYJBBGRKZCSX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Optimal catalyst for stereospecific and regioselective reaction of silacyclopropanes with carbonyl compounds.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franz, A.K. Woerpel, K.A.
Journal of the American Chemical Society, 121, 949-949 (1999)
J R Baker et al.
Organic letters, 9(1), 45-48 (2006-12-29)
[reaction: see text] A key structural feature of the Neocarzinostatin chromophore is a reactive epoxydiyne. We present here a new method for the preparation of epoxydiynes by the addition of an allenyl zinc bromide to a propargylic ketone.
Fernando Ferreira et al.
Nucleosides, nucleotides & nucleic acids, 24(5-7), 1009-1013 (2005-10-27)
An oligonucleotide protected with N-(trimethylsilyloxycarbonyl) (Teoc) and P-(trimethylsilylethanol) (Tse) groups was synthesized and deprotected by a single ZnBr2 treatment. Finally it was released from the solid support by cleavage of a disulfide linkage with TCEP. The olgonucleotide was obtained without
Biswajit Kalita et al.
Chemical communications (Cambridge, England), (36)(36), 4291-4293 (2008-09-20)
Benzylic and allylic hydrocarbons are selectively converted to the corresponding sulfonamides by a ZnBr2-H2O-catalyzed reaction with PhI=NTs; saturated adamantane is aminosulfonated at the tertiary C-H bond.
Yasuhiro Inui et al.
Chemical & pharmaceutical bulletin, 57(10), 1158-1160 (2009-10-06)
Asymmetric intermolecular hydroacylation between salicylaldehyde (1) and 1,5-hexadiene (2) using a combination of [RhCl(C(8)H(14))(2)](2) (0.10 eq), (S)-BINAP (0.10 eq), and ZnBr(2) (0.20 eq) afforded an enantiomerically enriched hydroacylated product iso-3 of 84% ee, along with an achiral product normal-3.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service