Skip to Content
Merck
All Photos(1)

Documents

450936

Sigma-Aldrich

Iron(II) chloride

AnhydroBeads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Ferrous chloride, Iron dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl2
CAS Number:
Molecular Weight:
126.75
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.99% trace metals basis

impurities

≤150.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

677 °C (lit.)

density

3.16 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Fe]Cl

InChI

1S/2ClH.Fe/h2*1H;/q;;+2/p-2

InChI key

NMCUIPGRVMDVDB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Iron(II) chloride, also known as ferrous chloride, is a paramagnetic solid with a high melting point. It has significant applications in solar cells, fuel cell catalysts, and semiconductors due to its unique properties. Additionally, it can also act as a reducing agent in redox reactions, which are crucial in many electrochemical processes.

Application

Iron(II) chloride can be used as:      
  • A doping agent in the fabrication of mixed organic-inorganic perovskite for high-efficiency perovskite solar cells. The incorporation of iron(II) chloride improve the overall efficiency of the solar cell.     
  • A precursor for synthesizing various iron(II) coordination complexes. These complexes are significant due to their unique optical and electronic properties, which are essential for various applications, including solar energy conversion and catalysis.     
  • As a precursor in the synthesis of iron-nitrogen-carbon (Fe–N–C) catalysts for proton exchange membrane fuel cells.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inorganic Chemistry, 37, 612-612 (1998)
Jiangfei Meng et al.
Journal of food science, 77(1), C8-14 (2011-12-21)
Wines made from 3 spine grape (Vitis davidii Foex) genotypes-Junzi 1# (JZ 1#), Junzi 2# (JZ 2#), and Liantang (LT)-and Cherokee rose (Rosa laevigata Michx., CR) were evaluated for their phenolics composition and antioxidant activities by several assays, including 2,2'-azino-bis-(3-ethylbenzothiazoline-6-sulfonic
Sofia Tresintsi et al.
Water research, 46(16), 5255-5267 (2012-07-25)
Various iron oxy-hydroxides were synthesized in a continuous flow kilogram-scale production reactor through the precipitation of FeSO(4) and FeCl(2) in the pH range 3-12 under intense oxidative conditions to serve as arsenic adsorbents. The selection of the optimum adsorbent and
Qinqin Xia et al.
The Journal of organic chemistry, 77(20), 9366-9373 (2012-10-03)
Iron-catalyzed direct C-N bond formation between azoles and amides is described. The oxidative coupling reactions of sp(3) C-H bonds adjacent to a nitrogen atom in amides and sulfonamides with the N-H bond in azoles proceeded smoothly in the presence of
Angéla Hajdú et al.
Colloids and surfaces. B, Biointerfaces, 94, 242-249 (2012-03-01)
Magnetite nanoparticles (MNPs) were prepared by alkaline hydrolysis of Fe(II) and Fe(III) chlorides. Adsorption of polyacrylic acid (PAA) on MNPs was measured at pH=6.5±0.3 and I=0.01 M (NaCl) to find the optimal PAA amount for MNP stabilization under physiological conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service