Skip to Content
Merck
All Photos(1)

Key Documents

434000

Sigma-Aldrich

Iron(II) bromide

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Ferrous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeBr2
CAS Number:
Molecular Weight:
215.65
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
beads
solubility:
ethanol: very soluble(lit.)

product line

AnhydroBeads

Quality Level

Assay

99.999% trace metals basis

form

beads

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

bp

934 °C (lit.)

mp

684 °C (lit.)

solubility

ethanol: very soluble(lit.)

density

4.63 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Br[Fe]Br

InChI

1S/2BrH.Fe/h2*1H;/q;;+2/p-2

InChI key

GYCHYNMREWYSKH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pavel Chábera et al.
Nature, 543(7647), 695-699 (2017-03-31)
Transition-metal complexes are used as photosensitizers, in light-emitting diodes, for biosensing and in photocatalysis. A key feature in these applications is excitation from the ground state to a charge-transfer state; the long charge-transfer-state lifetimes typical for complexes of ruthenium and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service