Skip to Content
Merck
All Photos(3)

Key Documents

417963

Sigma-Aldrich

Chitosan

from shrimp shells, practical grade

Synonym(s):

Deacetylated chitin, Poly(D-glucosamine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.23

biological source

shrimp shells

grade

practical grade

form

solid

mol wt

190000-375000 Da

viscosity

>200 cP, 1 wt. % in 1% acetic acid(20 °C, Brookfield)(lit.)

solubility

1 M acetic acid: 10 mg/mL

SMILES string

N([C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O)O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4N)O)O[C@@H]5O[C@@H]([C@H]([C@@H]([C@H]5N)O)O[C@@H]6O[C@@H]([C@H]([C@@H]([C@H]6N)O)O[C@@H]7O[C@@H]([C@H]([C@@H]([C@H]7N)O)O[C@@H]8O[C@@H]([C@H]([C@@H]([C@H]8N)O)O[C@@H]9O[C@@H]([C@H](

InChI

1S/C56H103N9O39/c1-87-56(86)65-28-38(84)46(19(10-74)96-55(28)104-45-18(9-73)95-49(27(64)37(45)83)97-39-12(3-67)88-47(85)20(57)31(39)77)103-54-26(63)36(82)44(17(8-72)94-54)102-53-25(62)35(81)43(16(7-71)93-53)101-52-24(61)34(80)42(15(6-70)92-52)100-51-23(60)33(79)41(14(5-69)91-51)99-50-22(59)32(78)40(13(4-68)90-50)98-48-21(58)30(76)29(75)11(2-66)89-48/h11-55,66-85H,2-10,57-64H2,1H3,(H,65,86)/t11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48+,49+,50+,51+,52+,53+,54+,55+/m1/s1

InChI key

FLASNYPZGWUPSU-SICDJOISSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Forms gels with multivalent anions. Gives clear solutions that dry to strong, clear films.

Features and Benefits

Biocompatible, antibacterial and environmentally friendly polyelectrolyte with a variety of applications including water treatment, chromatography, additives for cosmetics, textile treatment for antimicrobial activity, novel fibers for textiles, photographic papers, biodegradable films, biomedical devices, and microcapsule implants for controlled release in drug delivery.
Biocompatible, antibacterial and environmentally friendly polyelectrolyte.

Physical form

≥ 75% deacetylated

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arash Ghalayani Esfahani et al.
Journal of biomedical materials research. Part A, 107(7), 1455-1465 (2019-02-21)
One of the main challenges in the design of scaffolds for cortical bone regeneration is mimicking the highly oriented, hierarchical structure of the native tissue in an efficient, simple, and consistent way. As a possible solution to this challenge, positive
Alberto Busilacchi et al.
Carbohydrate polymers, 98(1), 665-676 (2013-08-31)
The idea of using chitosan as a functional delivery aid to support simultaneously PRP, stem cells and growth factors (GF) is associated with the intention to use morphogenic biomaterials to modulate the natural healing sequence in bone and other tissues.
Stephanie Supper et al.
Expert opinion on drug delivery, 11(2), 249-267 (2013-12-07)
Thermogelling chitosan (CS)/glycerophosphate (GP) solutions have been reported as a new type of parenteral in situ forming depot system. These free-flowing solutions at ambient temperature turn into semi-solid hydrogels after parenteral administration. Formulation parameters such as CS physico-chemical characteristics, CS/gelling
Liming Hu et al.
Nanoscale, 5(8), 3103-3111 (2013-03-22)
Within the past few years, chitosan-based drug delivery vehicles have become some of the most attractive to be studied. In contrast to all other polysaccharides, chitosan has demonstrated its unique characteristics for drug delivery platforms, including its active primary amino
J Pradeep Kumar et al.
Journal of biomedical nanotechnology, 10(6), 970-981 (2014-04-23)
The scaffolds for bone tissue engineering should be porous to harbor the growth of new tissue ingrowths, biodegradable with no toxic end products, and biocompatible with no cytotoxicity. In this study we report that Diopside (CaMgSi2O6) (Dp) particles can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service