Skip to Content
Merck
All Photos(1)

Key Documents

446955

Sigma-Aldrich

Allyltrimethoxysilane

95%

Synonym(s):

Trimethoxy-2-propen-1-ylsilane, trimethoxy-2-propenylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2Si(OCH3)3
CAS Number:
Molecular Weight:
162.26
Beilstein:
2350745
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.405 (lit.)

bp

146-148 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CC=C)(OC)OC

InChI

1S/C6H14O3Si/c1-5-6-10(7-2,8-3)9-4/h5H,1,6H2,2-4H3

InChI key

LFRDHGNFBLIJIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allyltrimethoxysilane is an allylating reagent that can be used for the allylation of carbonyl compounds such as aldehydes, ketones, and imines. Homoallylic alcohols and amines are obtained via the C-C bond forming reaction. It can also be used to synthesize homoallylic α-branched amines from aromatic and aliphatic aldehyde hydrazones, and ketone hydrazones.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allyltrimethoxysilane addition to N-acylhydrazones: Two catalytic methods employing CuCl and fluoride
Ding H and Friestad GK
Synthesis, 2004(13), 2216-2221 (2004)
Shingo Yamasaki et al.
Journal of the American Chemical Society, 124(23), 6536-6537 (2002-06-06)
A general and mild catalytic allylation of carbonyl compounds, applicable to aldehydes, ketones, and imines is developed using allyltrimethoxysilane as the allylating reagent. The reaction proceeds smoothly with 1-10 mol % of CuCl and TBAT in THF at ambient temperature.
Hisashi Yamamoto et al.
Chemistry, an Asian journal, 2(6), 692-698 (2007-05-25)
Recently, there have been some reports on the use of allyltrimethoxysilane for enantioselective allylation with various metal fluorides or a combination of chiral complex and fluoride anion. These reactions can be applied not only to aldehydes but also to ketones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service