Skip to Content
Merck
All Photos(1)

Key Documents

446440

Sigma-Aldrich

5-Norbornene-2-carboxylic acid, mixture of endo and exo, predominantly endo

98%

Synonym(s):

2-Norbornene-5-carboxylic acid, Bicyclo[2.2.1]hept-2-ene-5-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O2
CAS Number:
Molecular Weight:
138.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.494 (lit.)

bp

136-138 °C/14 mmHg (lit.)

density

1.129 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)C1C[C@H]2C[C@@H]1C=C2

InChI

1S/C8H10O2/c9-8(10)7-4-5-1-2-6(7)3-5/h1-2,5-7H,3-4H2,(H,9,10)/t5-,6+,7?/m1/s1

InChI key

FYGUSUBEMUKACF-JEAXJGTLSA-N

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of well-defined graft copolymers via coupled living anionic polymerization and living ROMP.
Rizmi ACM, et al.
Polymer, 39(25), 6605-6610 (1998)
A Convenient Practical Method for the Preparation of (-)-(1 S, 2 S)-5-Norbornene-2-carboxylic Acid, Incorporating Efficient Recovery of the Chiral Auxiliary d-Pantolactone.
Chang H, et al.
Organic Process Research & Development, 3(4), 289-291 (1999)
Controlled Synthesis of Reactive Polymeric Architectures Using 5-Norbornene-2-carboxylic Acid Pentafluorophenyl Ester.
Vogel N and Theato P.
Macromolecular Symposia, 249(1) (2007)
Benjamin D Fairbanks et al.
Advanced materials (Deerfield Beach, Fla.), 21(48), 5005-5010 (2009-12-28)
Step-growth, radically mediated thiol-norbornene photopolymerization is used to create versatile, stimuli-responsive poly(ethylene glycol)-co-peptide hydrogels The reaction is cytocompatible and allows for the encapsulation of human mesenchymal stem cells with a viability greater than 95%. Cellular spreading is dictated via three-dimensional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service