247634
cis-5-Norbornene-endo-2,3-dicarboxylic anhydride
99%
Synonym(s):
cis-endo-5-Norbornene-2,3-dicarboxylic anhydride, cis-endo-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C9H8O3
CAS Number:
Molecular Weight:
164.16
Beilstein:
83657
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99%
mp
165-167 °C (lit.)
SMILES string
[H][C@@]12C3CC(C=C3)[C@]1([H])C(=O)OC2=O
InChI
1S/C9H8O3/c10-8-6-4-1-2-5(3-4)7(6)9(11)12-8/h1-2,4-7H,3H2/t4-,5+,6-,7+
InChI key
KNDQHSIWLOJIGP-UMRXKNAASA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chao-Bin Yeh et al.
European journal of medicinal chemistry, 45(9), 3981-3985 (2010-08-10)
Previous research indicates that cantharidin, norcantharidin and their analogues exhibit anticancer activity due to their inhibition of cancer cell lines such as HL60, HT29 and L1210. The anticancer activities of cantharidin, norcantharidin and their analogues involve the suppression of serine/threonine
Kendra Fein et al.
Carbohydrate polymers, 250, 117001-117001 (2020-10-15)
Cellulose nanofibrils (CNF) coatings are excellent grease barriers for biodegradable packaging. However, barrier properties are moisture sensitive, so hydrophobic components such as latexes or polymers are needed to impart moisture resistance, but incompatibility leads to poor dispersion. In this work
Malcolm Driffield et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(6), 1200-1213 (2018-02-24)
The polymeric coating used in metal packaging such as cans for foods and beverages may contain residual amounts of monomers used in the production of the coating, as well as unreacted linear and cyclic oligomers. Traditionally, although designed for use
A sequential 3D bioprinting and orthogonal bioconjugation approach for precision tissue engineering.
Claire Yu et al.
Biomaterials, 258, 120294-120294 (2020-08-18)
Recent advances in 3D bioprinting have transformed the tissue engineering landscape by enabling the controlled placement of cells, biomaterials, and bioactive agents for the biofabrication of living tissues and organs. However, the application of 3D bioprinting is limited by the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service