All Photos(1)
About This Item
Linear Formula:
H2NCOCH=CHCO2H
CAS Number:
Molecular Weight:
115.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
solid
Quality Level
mp
158-161 °C (lit.)
functional group
amide
carboxylic acid
SMILES string
[H]\C(=C(/[H])C(O)=O)C(N)=O
InChI
1S/C4H5NO3/c5-3(6)1-2-4(7)8/h1-2H,(H2,5,6)(H,7,8)/b2-1-
InChI key
FSQQTNAZHBEJLS-UPHRSURJSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Maleamic and citraconamic acids, methyl esters, and imides.
MEHTA NB, et al.
The Journal of Organic Chemistry, 25(6), 1012-1015 (1960)
Meifang Huang et al.
International journal of biological macromolecules, 36(1-2), 98-102 (2005-06-14)
N-maleamic acid-chitosan was synthesized and characterized by Fourier transform infrared spectra analysis (FT-IR) and 1H NMR. The graft copolymerization of N-maleamic acid-chitosan and butyl acrylate (BA) in acetic acid aqueous solution was investigated, using the gamma-ray of 60Co gamma-irradiation method.
Juan Rodrigues et al.
Pharmaceutical research, 28(4), 907-919 (2010-12-25)
The objective of this study was to investigate the effect of new 4-maleamic acid and 4-maleamide peptidyl chalcone derivatives against human prostate cancer in vitro and in vivo. From a series of 21 chalcones, the effects of the three best
Yoshihiro Miyaji et al.
Pharmaceutical research, 29(11), 3143-3155 (2012-06-26)
Recombinant osteoprotegerin (OPG) has been proven to be useful for treating various bone disorders such as osteoporosis. To improve its in vivo pharmacological effect, OPG was conjugated to novel comb-shaped co-polymers of polyethylene glycol (PEG) allylmethylether and maleamic acid (poly(PEG)
Shengzu Zhang et al.
Journal of separation science, 31(21), 3782-3787 (2008-10-18)
The polymerizable gelator N-octadecyl maleamic acid (ODMA) can self-assemble in selected polymerizable organic solvents, such as 2-hydroxyethyl methacrylate (HEMA) and methylacrylic acid (MAA) to form thermally stable polymerizable organogels. A mixture consisting of HEMA and MAA as the monomer and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service