Skip to Content
Merck
All Photos(1)

Documents

442291

Sigma-Aldrich

Octadecylsilane

97%

Synonym(s):

Octadecyltrihydridosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17SiH3
CAS Number:
Molecular Weight:
284.60
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

195 °C/15 mmHg (lit.)

mp

29 °C (lit.)

density

0.795 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC[SiH3]

InChI

1S/C18H40Si/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-18H2,1,19H3

InChI key

YTJSFYQNRXLOIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octadecylsilane is an organosilicon compound that consists of a long-chain hydrocarbon (18 carbons) bonded to a silicon atom through a carbon-silicon bond. It is widely used in various applications due to its high surface activity, making it an excellent adsorbent for non-polar organic compounds. It is also used as a surface modifier or coating agent.

Application

Octadecylsilane can be used as a modifying agent for montmorillonite clay nanoparticles, which are used as a filler material in the synthesis of poly(methyl methacrylate) (PMMA)/silylated montmorillonite composites.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thanh-Thien Tran-Lam et al.
Foods (Basel, Switzerland), 7(12) (2018-12-14)
Polycyclic aromatic hydrocarbons (PAHs)-a large group of organic compounds-are extremely hazardous to human health. In this study, the 198 samples from six groups of daily food products in the Hanoi metropolitan area were collected and prepared by the QuEChERS sample
Mohammad Arafat et al.
Pharmaceutics, 13(1) (2020-12-31)
Self-expanding metal stents (SEMSs) are currently the gold standard for the localised management of malignant gastrointestinal (GI) stenosis and/or obstructions. Despite encouraging clinical success, in-stent restenosis caused by tumour growth is a significant challenge. Incorporating chemotherapeutic drugs into GI stents
Takumi Tomono et al.
Journal of pharmaceutical sciences, 106(9), 2642-2649 (2017-03-23)
Overexpression and activation of P-glycoprotein (P-gp), which mediates efflux transport of various anticancer drugs in cancer cells, is associated with multidrug resistance (MDR). On the other hand, malignant cancer cells frequently undergo epithelial-to-mesenchymal transition (EMT), thereby acquiring high migratory mobility
Marcelo Delmar Cantú et al.
Analytical and bioanalytical chemistry, 386(2), 256-263 (2006-08-10)
Simple, sensitive, and reproducible off-line solid-phase microextraction and liquid chromatography (SPME/LC) methods are described for the determination of seven anticonvulsants and tricyclic antidepressants in human plasma. Factorial design and simplex methodology were applied in the optimization of the SPME procedure
Tomokazu Hata et al.
PloS one, 12(7), e0180745-e0180745 (2017-07-07)
Gut lumen serotonin (5-hydroxytryptamine: 5-HT) contributes to several gastrointestinal functions such as peristaltic reflexes. 5-HT is released from enterochromaffin (EC) cells in response to a number of stimuli, including signals from the gut microbiota. However, the specific mechanism by which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service