Skip to Content
Merck
All Photos(1)

Documents

440248

Sigma-Aldrich

Dichloromethylsilane

≥97%

Synonym(s):

Methyldichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SiHCl2
CAS Number:
Molecular Weight:
115.03
Beilstein:
1071194
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

vapor density

4 (vs air)

Quality Level

vapor pressure

6.79 psi ( 20 °C)

Assay

≥97%

form

liquid

autoignition temp.

471 °F

expl. lim.

>55 %

refractive index

n20/D 1.398 (lit.)

bp

41 °C (lit.)

mp

−93 °C (lit.)

density

1.105 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[SiH](Cl)Cl

InChI

1S/CH4Cl2Si/c1-4(2)3/h4H,1H3

InChI key

NWKBSEBOBPHMKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyldichlorosilane(DMDCS) is a tetrahedral organosilicon compound. It is used as a silane coupling agent to synthesis the hydrophilic and hydrophobic silica nanoparticles. It is also used in the synthesis of polysilanes, which are precursors to silicon carbide.

Application


  • A General and Selective Synthesis of Methylmonochlorosilanes from Di-, Tri-, and Tetrachlorosilanes - Discusses a method for synthesizing Methylmonochlorosilanes, showing potential for varied applications in chemical synthesis (Y Naganawa et al., 2020).

  • Polymerization of methylsilylenes into polymethylsilanes or polycarbosilanes after dechlorination of dichloromethylsilanes. - Investigates the polymerization of methylsilylenes, offering insights into the production of polymethylsilanes or polycarbosilanes from Dichloromethylsilanes (Y Tian et al., 2016).

  • Synthesis of low viscosity of polymethylhydrosiloxane using monomer of dichloromethylsilane - Focuses on producing low-viscosity polymethylhydrosiloxane through hydrolysis-condensation of Dichloromethylsilane, significant for various industrial applications (VF Arini et al., 2022).


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - Water-react 3

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-18.4 °F

Flash Point(C)

-28 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Multihydroxy-functional polysilanes via an acetal protecting group strategy
Reuss VS and Frey H
Macromolecules, 43(20) (2010)
Jing Li et al.
Journal of cellular physiology, 235(9), 6139-6153 (2020-02-06)
Atherosclerosis is one of the leading causes of morbidity and mortality, mainly due to the immune response triggered by the recruitment of monocytes/macrophages in the artery wall. Accumulating evidence have shown that matrix stiffness and oxidized low-density lipoproteins (ox-LDL) play
K Kim et al.
Journal of biomedical materials research, 52(4), 836-840 (2000-10-18)
In order to develop a versatile model for blood-compatible materials, we studied morphology and platelet adhesion of a dipalmitoyl-phosphatidylcholine (DPPC)/dipalmitoyl-phosphatidylethanolamine-polyethylene glycol (PEG lipid) mixed monolayer. This monolayer, which mimics the cell membrane structure, consists of two heterogeneous layers, that is
S G Khaspekova et al.
Biochemistry. Biokhimiia, 81(2), 114-121 (2016-06-05)
Activity of tissue factor (TF) in membrane microparticles (MPs) produced in vitro by endothelial cells (ECs), monocytes, THP-1 monocytic cells, granulocytes, and platelets was investigated. ECs were isolated from human umbilical vein, and monocytes, granulocytes, and platelets - from the
Jaleh Varshosaz et al.
Drug delivery, 11(1), 53-58 (2004-06-01)
Hydroxyethylmethacrylate/methacrylic acid copolymer cross-linked with ethylenglycol dimethacrylate was prepared by a bulk free radical polymerization method. The permeability studies of this pH-sensitive hydrogel to drugs with different water solubilities showed a water-content dependent diffusion or pore mechanism for ephedrine HCl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service