Skip to Content
Merck
All Photos(1)

Key Documents

40140

Sigma-Aldrich

Dichlorodimethylsilane

≥98.5% (GC)

Synonym(s):

DMDCS, Dimethyldichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SiCl2
CAS Number:
Molecular Weight:
129.06
Beilstein:
605287
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (GC)

form

liquid

refractive index

n20/D 1.404 (lit.)

bp

70 °C (lit.)

mp

−76 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(Cl)Cl

InChI

1S/C2H6Cl2Si/c1-5(2,3)4/h1-2H3

InChI key

LIKFHECYJZWXFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dichlorodimethylsilane (DDMS) can be used as a silylating agent for the silylation of Pd/SiO2 catalysts to enhance the catalytic activity for the selective hydrogenation of aromatic ketones.
It is also employed as a reagent in the synthesis of hydrophilic and hydrophobic silica nanoparticles and amides from unprotected amino acids.

Other Notes

Silylating agent for diols, diamines, etc.; the protection is very sensitive to hydrolysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuning the support adsorption properties of Pd/SiO2 by silylation to improve the selective hydrogenation of aromatic ketones
Quintanilla A, et al.
J. Catal., 257(1), 55-63 (2008)
Synthesis of amides from unprotected amino acids by a simultaneous protection-activation strategy using dichlorodialkyl silanes
Van Leeuwen SH, et al.
Tetrahedron Letters, 43(50), 9203-9207 (2002)
Water-repellent acylated and silylated wood samples and their surface analytical characterization
Mohammed-Ziegler I, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 319(1-3), 204-212 (2008)
Dichlorodimethylsilane mediated one-step synthesis of hydrophilic and hydrophobic silica nanoparticles
Ebrahimi F, et al.
Advanced Powder Technology, 28(3), 932-937 (2017)
J. Heberle et al.
Silylating Agents, 2nd ed. (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service