Skip to Content
Merck
All Photos(2)

Key Documents

438472

Sigma-Aldrich

Potassium borohydride

99.9% trace metals basis

Synonym(s):

Potassium tetrahydroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBH4
CAS Number:
Molecular Weight:
53.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: reductant

impurities

≤1% Na

mp

500 °C (dec.) (lit.)

density

1.18 g/mL at 25 °C (lit.)

SMILES string

[K+].[H][B-]([H])([H])[H]

InChI

1S/BH4.K/h1H4;/q-1;+1

InChI key

ICRGAIPBTSPUEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenli Zhu et al.
Analytica chimica acta, 607(2), 136-141 (2008-01-15)
A new atomizer based on atmospheric pressure dielectric barrier discharge (DBD) plasma was specially designed for atomic fluorescence spectrometry (AFS) in order to be applied to the measurement of arsenic. The characteristics of the DBD atomizer and the effects of
Chuan Zhang et al.
Analytica chimica acta, 652(1-2), 143-147 (2009-09-30)
Room-temperature ionic liquid in combination with sodium diethyldithiocarbamate (DDTC) was used to synergetically improve the chemical vapor generation (CVG) of nickel. Volatile species of nickel were effectively generated through reduction of acidified analyte solution with KBH(4) in the presence of
A P Seddon et al.
The Journal of biological chemistry, 261(25), 11538-11543 (1986-09-05)
Bacterial 5-oxoprolinase is composed of two protein components: Component A, which catalyzes 5-oxoproline-dependent ATP-hydrolysis and Component B, which couples the hydrolysis of ATP with the decyclization of 5-oxoproline to form glutamate (Seddon, A. P., Li, L., and Meister, A. (1984)
Jesus R Medina et al.
The Journal of organic chemistry, 68(12), 4631-4642 (2003-06-07)
Representative organoborane mixtures were quantitatively converted to their borohydrides through their reaction with activated KH (KH), permitting their detailed analysis by (11)B NMR. Through the treatment of commercial KH with a THF solution of lithium aluminum hydride (LAH), a dramatic
Feng Li et al.
Journal of chromatography. A, 1081(2), 232-237 (2005-07-26)
A novel method for speciation analysis of inorganic arsenic was developed by on-line hyphenating microchip capillary electrophoresis (chip-CE) with hydride generation atomic fluorescence spectrometry (HG-AFS). Baseline separation of As(III) and As(V) was achieved within 54 s by the chip-CE in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service