Skip to Content
Merck
All Photos(1)

Key Documents

433608

Sigma-Aldrich

Strontium hydroxide

94%

Synonym(s):

Strontium dihydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sr(OH)2
CAS Number:
Molecular Weight:
121.63
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks

Quality Level

Assay

94%

form

powder and chunks

impurities

3% SrCO3

SMILES string

O[Sr]O

InChI

1S/2H2O.Sr/h2*1H2;/q;;+2/p-2

InChI key

UUCCCPNEFXQJEL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zh G Mustafina et al.
Vestnik oftalmologii, 115(5), 28-29 (1999-12-03)
Strontium hydroxide Sr(OH)2 has been identified in tumor tissues of patients with melanoma of the vascular coat of the eye and in metastatic tumors to the vascular coat of the eye by x-ray structural analysis. Normal vascular coat of the
H Arcis et al.
Physical chemistry chemical physics : PCCP, 16(33), 17688-17704 (2014-07-18)
Frequency-dependent electrical conductivities of solutions of aqueous strontium hydroxide and strontium chloride have been measured from T = 295 K to T = 625 K at p = 20 MPa, over a very wide range of ionic strength (3 ×
Histochemical analysis of the dogs' dental pulp after pulp capping with calcium, barium, and strontium hydroxides.
R Holland et al.
Journal of endodontics, 8(10), 444-447 (1982-10-01)
Mohammad Amin Alavi et al.
Ultrasonics sonochemistry, 17(1), 132-138 (2009-06-09)
The Sr(OH)(2) and SrCO(3) nanostructures were synthesized by reaction of strontium(II) acetate and sodium hydroxide or tetramethylammonium hydroxide (TMAH) via ultrasonic method. Reaction conditions, such as the concentration of the Sr(2+) ion, aging time, power of the ultrasonic device and
Chang-Min Yoon et al.
ACS applied materials & interfaces, 7(34), 18977-18984 (2015-08-13)
A series of alkaline earth metal-doped hollow SiO2/TiO2 spheres (EM-HST) are prepared as electrorheological (ER) materials via sonication-mediated etching method with various alkaline earth metal hydroxides as the etchant. The EM-HST spheres are assessed to determine how their hollow interior

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service