Skip to Content
Merck
All Photos(2)

Key Documents

204498

Sigma-Aldrich

Strontium nitrate

99.995% trace metals basis

Synonym(s):

Strontium dinitrate, Strontium(II) nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sr(NO3)2
CAS Number:
Molecular Weight:
211.63
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38060313
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps

Quality Level

Assay

99.995% trace metals basis

form

crystals and lumps

impurities

≤55.0 ppm Trace Metal Analysis

pH

5.0-7.0 (25 °C)

mp

570 °C (lit.)

SMILES string

[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Sr/c2*2-1(3)4;/q2*-1;+2

InChI key

DHEQXMRUPNDRPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Strontium nitrate may be used as one of the starting materials for the preparation of calcium substituted strontium hexaferrite nanoparticles and strontium ferrite (SrFe12O19) particles by the chemical co-precipitation method. The product may also be used for the synthesis of α- and β-(Sr0.97Eu0.03)2SiO4 phosphors for near UV light emitting diodes.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure dependent luminescence characterization of green?yellow emitting Sr2SiO4:Eu2+ phosphors for near UV LEDs
Han JK, et al.
Journal of Luminescence, 132.1, 106-109 (2012)
Influence of surfactants on co-precipitation synthesis of strontium ferrite
Lu H, et al.
J. Alloy Compounds, 509.41, 10127-10131 (2011)
Physical, electrical and dielectric properties of Ca-substituted strontium hexaferrite (SrFe 12 O 19) nanoparticles synthesized by co-precipitation method
Iqbal MJ, et al.
Journal of magnetism and magnetic materials, 322.13, 1720-1726 (2010)
G S Hahn
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 25(9), 689-694 (1999-09-22)
Irritant contact dermatitis occurs following topical exposure to many chemicals found in cosmetics, personal care products, drugs, and during occupational exposure. Signs and symptoms may include sensory irritation (sting, burn, and/or itch), erythema, edema, and vesiculation. In an attempt to
H Zhai et al.
Contact dermatitis, 42(2), 98-100 (2000-03-07)
Skin care products are complex formulations that may cause sensory irritation symptoms, characterized by stinging, burning, and itching. Substances capable of counteracting sensory irritation are of great practical interest. Strontium salts have been demonstrated to inhibit sensory irritation and inflammation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service