Skip to Content
Merck
All Photos(2)

Key Documents

432504

Sigma-Aldrich

Sodium amide

greener alternative

95%

Synonym(s):

Sodamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNH2
CAS Number:
Molecular Weight:
39.01
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

400 °C (lit.)

mp

210 °C (lit.)

greener alternative category

SMILES string

N[Na]

InChI

1S/H2N.Na/h1H2;/q-1;+1

InChI key

ODZPKZBBUMBTMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Reagent for synthesis of:
Allylic amines and amides
Antibacterials

Reagent for:
Aggregative activation and heterocyclic chemistry
Phenylation with diphenyliodonium chloride
Sodium amide (NH2Na) (95%) can be used for a variety of applications such as:
  • production of a hydrogen storage system
  • adsorption of CO2
  • fabrication of fuel cell

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Bakavoli, et al.,
Journal of Heterocyclic Chemistry, 48, 149-152 (2010)
Hydrogen production from ammonia using sodium amide
David WIF, et al.
Journal of the American Chemical Society, 136(38), 13082-13085 (2014)
Thermal decomposition kinetics of light-weight composite NaNH2-NaBH4 hydrogen storage materials for fuel cells
Bai Y, et al.
International Journal of Hydrogen Energy, 37(17), 12973-12979 (2012)
Christian Bukovec et al.
Organic & biomolecular chemistry, 9(8), 2743-2750 (2011-03-08)
Stannylated allylic carbonates are suitable substrates for Pd-catalyzed allylic aminations. In DMF and with [allylPdCl](2) as catalyst the stannylated allyl amines formed can be directly coupled with electrophiles according to the Stille protocol, giving rise to highly functionalized building blocks
Highly efficient CO2 adsorption by nitrogen-doped porous carbons synthesized with low-temperature sodium amide activation
Wang L, et al.
Carbon, 130(16), 31-40 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service