Skip to Content
Merck
All Photos(1)

Key Documents

421626

Sigma-Aldrich

(S)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
Beilstein:
4842199
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

optical activity

[α]20/D −168°, c = 1.8 in 1.4 M NaOH

reaction suitability

reaction type: solution phase peptide synthesis

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)[C@@H]1Cc2ccccc2CN1

InChI

1S/C10H11NO2/c12-10(13)9-5-7-3-1-2-4-8(7)6-11-9/h1-4,9,11H,5-6H2,(H,12,13)/t9-/m0/s1

InChI key

BWKMGYQJPOAASG-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Li et al.
Nanomedicine : nanotechnology, biology, and medicine, 8(7), 1216-1222 (2012-01-18)
The modification of 3S-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid (THIQA) with β-cyclodextrin (β-CD) provides an oral antithrombotic agent, 6-(3'S-isoquinoline-3'-carboxylaminoethylamino)-6-deoxy-β-CD (THIQA-β-CD). In aqueous solution THIQA-β-CD undergoes intermolecular inclusion complexation and forms pH-dependent nanostructures. The morphological feature of THIQA-β-CD is a nanocloud consisting of numerous particles
Bradford H Hirth et al.
Bioorganic & medicinal chemistry letters, 15(8), 2087-2091 (2005-04-06)
A series of 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid diamides that increase chloride transport in cells expressing mutant cystic fibrosis transmembrane conductance regulator (CFTR) protein has been identified from our compound library. Analoging efforts and the resulting structure-activity relationships uncovered are detailed. Compound potency
Wei-Jie Fang et al.
Biopolymers, 96(1), 97-102 (2010-06-19)
Peptides containing N-methylamino acids can exhibit improved pharmacodynamic and pharmacokinetic profiles compared to nonmethylated peptides, and therefore interest in these N-methylated peptides has been increasing in recent years. Arodyn (Ac[Phe¹,²,³,Arg⁴,D-Ala⁸]Dyn A(1-11)NH₂) is an acetylated dynorphin A(Dyn A) analog that is
D Pagé et al.
Journal of medicinal chemistry, 44(15), 2387-2390 (2001-07-13)
The design, synthesis and pharmacological evaluation of a novel class of Dmt-Tic dipeptide analogues are described. These resulting analogues bearing different C-terminal functionalities were found to bind to the human delta receptor with high affinity. One specific class of dipeptides
R Guerrini et al.
Bioorganic & medicinal chemistry, 6(1), 57-62 (1998-03-21)
Substitution of 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid (Tic) in place of Gly2 in dynorphin A-(1-13)-NH2 and -(1-11)-NH2 (DYN) analogues (1 and 2) decreased the affinity to the kappa, delta, and mu receptors, and kappa selectivity. The analogue [D-Ala2, des-Gly3]DYN (4), a chimera between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service