Skip to Content
Merck
All Photos(1)

Key Documents

420581

Sigma-Aldrich

Glycolic acid solution

high purity, 70 wt. % in H2O

Synonym(s):

Hydroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2COOH
CAS Number:
Molecular Weight:
76.05
Beilstein:
1209322
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021303
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

quality

high purity

concentration

63-72% (by NaOH, titration)
70 wt. % in H2O

refractive index

n20/D 1.408

bp

112 °C

density

1.25 g/mL at 25 °C

functional group

carboxylic acid
hydroxyl

SMILES string

OCC(O)=O

InChI

1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)

InChI key

AEMRFAOFKBGASW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glycolic acid solution may be employed to evaluate the efficacy of glycolic peels treatment for all types of acne.[1]

Legal Information

DuPont product

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation and reduction of glycolic and glyoxylic acids in plants. I. Glycolic and oxidase.
I ZELITCH et al.
The Journal of biological chemistry, 201(2), 707-718 (1953-04-01)
N Wang et al.
Journal of biomaterials science. Polymer edition, 8(12), 905-917 (1997-01-01)
A series of oligomers or low molecular weight polymers of lactic and/or glycolic acid has been synthesized with different molar ratios of lactic to glycolic acid. These oligomers have been characterized with respect to oligomer composition, molecular weight, intrinsic viscosity
L Atzori et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 12(2), 119-122 (1999-05-27)
Glycolic acid chemical peels have been widely accepted as a useful modality in many cutaneous conditions characterized by abnormalities of keratinization. The aim of this study is to evaluate the use of glycolic peels in the main clinical forms of
F F Becker et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 22(5), 463-465 (1996-05-01)
Seventy percent glycolic acid solutions are being commonly used as superficial chemical peeling agents. The pH of these solutions ranges from 0.08 to 2.75. The histologic effects of these various pH solutions on human skin have not been studied. The
Ingela Bjurhager et al.
Biomacromolecules, 13(8), 2521-2527 (2012-07-10)
In 1628, the Swedish warship Vasa capsized on her maiden voyage and sank in the Stockholm harbor. The ship was recovered in 1961 and, after polyethylene glycol (PEG) impregnation, it was displayed in the Vasa museum. Chemical investigations of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service