Skip to Content
Merck
All Photos(1)

Key Documents

L6661

Sigma-Aldrich

Lactic acid

meets USP testing specifications

Synonym(s):

DL-Lactic acid, 2-Hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)COOH
CAS Number:
Molecular Weight:
90.08
Beilstein:
1209341
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Agency

USP/NF
meets USP testing specifications

Assay

88.0-92.0%

form

viscous liquid

refractive index

n20/D 1.425 (lit.)

pH

2

bp

122 °C/15 mmHg (lit.)

density

1.209 g/mL at 25 °C (lit.)

cation traces

heavy metals: ≤0.001%

application(s)

pharmaceutical (small molecule)

storage temp.

room temp

SMILES string

CC(O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

In animals, lactic acid is a metabolic compound produced by proliferating cells and during anaerobic conditions such as strenuous exercise. Lactic acid can be oxidized back to pyruvate or converted to glucose via gluconeogenesis. Lactic acid is preferentially metabolized by neurons in several mammal species and during early brain development.

Other Notes

A concentrated solution of lactic acid is typically a mixture of lactic acid lactate and lactic acid. The concentration of this solution is approximately 90% (w/w).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conversion of biomass to 1, 2-propanediol by selective catalytic hydrogenation of lactic acid over silica-supported copper.
Cortright RD, et al.
Applied Catalysis. B, Environmental, 39(4), 353-359 (2002)
A literature review of poly (lactic acid).
Garlotta D.
Journal of Polymers and the Environment, 9(2), 63-84 (2001)
Kwon et al.
Enzyme and microbial technology, 26(2-4), 209-215 (2000-02-26)
Batch fermentation studies were performed to evaluate the potentials of a complex nitrogen source, soybean, as an alternative to yeast extract for the economical production of lactic acid by Lactobacillus rhamnosus. An enzyme-hydrolysate of soybean meal, Soytone, with an adequate
Mohamed Ali Abdel-Rahman et al.
Biotechnology advances, 31(6), 877-902 (2013-04-30)
Fermentative production of optically pure lactic acid has roused interest among researchers in recent years due to its high potential for applications in a wide range of fields. More specifically, the sharp increase in manufacturing of biodegradable polylactic acid (PLA)
Eileen F O' Shea et al.
Current opinion in biotechnology, 24(2), 130-134 (2013-01-23)
Lactic acid bacteria (LAB) produce a wide variety of antimicrobial peptides (bacteriocins) which contribute to the safety and preservation of fermented foods. This review discusses strategies that have been or could be employed to further enhance the commercial application of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service