Skip to Content
Merck
All Photos(1)

Key Documents

42000601

Sigma-Aldrich

PRONOVA® UP VLVM

very low viscosity sodium alginate, viscosity <20 mPa.s, 1 %(20 °C)(low), mannuronate monomer units ≥50 %

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.25

Quality Level

form

powder

mol wt

Mw <75 kDa (approximate)

composition

mannuronate monomer units, ≥50%
G/M ratio, ≥1.5

impurities

≤100 CFU/g Total viable count
≤100 EU/g Endotoxins

pH

5.5-8.5 (20 °C, 1% in water)

viscosity

<20 mPa.s, 1 %(20 °C) (
low)

General description

PRONOVA UP VLVM is a very low viscosity (less than 20 mPas) sodium alginate where minimum 50% of the monomer units are mannuronate.

Application

Examples of Pronova Alginates applications include: encapsulation of living cells and therapeutic proteins for cell therapy medications, cryoprotectant, bone putty binder for tissue engineering, 3D bioprinting, foams and cell tissue scaffolds, hydrogels, advanced drug delivery and implants, wound management, and medical devices.

Legal Information

Product of Dupont Nutrition USA, Inc.
PRONOVA is a registered trademark of International N&H USA, Inc.

Disclaimer

NOT FOR USE IN HUMANS

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service