Skip to Content
Merck
All Photos(1)

Key Documents

4270701

Sigma-Aldrich

Novatach LVM GRGDSP

GRGDSP-peptide coupled high M sodium alginate, mannuronate acid content ≥50 %

Synonym(s):

Peptide coupled alginate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

form

powder

composition

mannuronate acid content, ≥50%

impurities

≤100 EU/g Endotoxins

pH

5.5-8.5 (20 °C, 1% in water)

General description

Modified sodium alginate, grafted with RGD peptide sequences, Novatach Peptide-coupled UP Alginates are designed to facilitate and promote interaction between cells and alginate-based scaffolds and matrices. They are useful when cell-to-matrix interactions are beneficial. The GRGDSP peptide sequence is a common example of such adhesion ligand used for MSC stem cells. The peptide alginate is available with four different alginate backbones giving flexibility to match hydrogel properties in your application.

Application

Novatach Peptide-coupled UP Alginates: initiator for biological interactions between alginate hydrogels and cells, encapsulation of living cells and therapeutic proteins for cell therapy medications, 3D bioprinting, foams and cell tissue scaffolds, hydrogels, advanced drug delivery, etc.

Legal Information

Product of Dupont Nutrition USA, Inc.
NOVATACH is a trademark of Dupont Nutrition USA, Inc.

Disclaimer

NOT FOR USE IN HUMANS

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service