Skip to Content
Merck
All Photos(2)

Key Documents

413283

Sigma-Aldrich

1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene

99%

Synonym(s):

2,4,6-Tris(3,5-di-tert -butyl-4-hydroxybenzyl)mesitylene, 3,3′,3′′,5,5′,5′′-Hexa-tert -butyl-α,α′,α′′-(mesitylene-2,4,6-triyl)tri-p -cresol, 4,4′,4′′-[(2,4,6-Trimethyl-1,3,5-benzenetriyl)tris(methylene)]tris[2,6-bis(1,1-dimethylethyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6[CH2C6H2[C(CH3)3]2OH]3
CAS Number:
Molecular Weight:
775.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder

mp

248-250 °C (lit.)

SMILES string

Cc1c(Cc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)c(C)c(Cc3cc(c(O)c(c3)C(C)(C)C)C(C)(C)C)c(C)c1Cc4cc(c(O)c(c4)C(C)(C)C)C(C)(C)C

InChI

1S/C54H78O3/c1-31-37(22-34-25-40(49(4,5)6)46(55)41(26-34)50(7,8)9)32(2)39(24-36-29-44(53(16,17)18)48(57)45(30-36)54(19,20)21)33(3)38(31)23-35-27-42(51(10,11)12)47(56)43(28-35)52(13,14)15/h25-30,55-57H,22-24H2,1-21H3

InChI key

VSAWBBYYMBQKIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene(HEMA) is an antioxidant that can be used to prevent the oxidative degradation of plastics.

Application

HEMA can be used as an antioxidant in the polycondensation of poly(isosorbide maleate-co-succinate) and poly(isosorbide maleate) to prepare biobased based unsaturated polyester. It may also be used in the preparation of water soluble polyesters which can potentially be used as in drug delivery and industrial coatings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

609.8 °F

Flash Point(C)

321 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unsaturated, biobased polyesters and their cross-linking via radical copolymerization.
Jasinska L and Koning CE
Journal of Polymer Science Part A: Polymer Chemistry, 48(13), 2885-2895 (2010)
Waterborne polyesters partially based on renewable resources.
Jasinska L and Koning CE
Journal of Polymer Science Part A: Polymer Chemistry, 48(24), 5907-5915 (2010)
Identification of antioxidant and ultraviolet light stabilizing additives in plastics by liquid chromatography/mass spectrometry.
Vargo JD and Olson KL
Analytical Chemistry, 57(3), 672-675 (1985)
Simultaneous determination of antioxidants and ultraviolet absorbers by ultra-performance liquid chromatography in food simulants.
Li C, et al.
Food Analytical Methods, 7(9), 1755-1762 (2014)
E Dreassi et al.
Food additives and contaminants, 15(4), 466-472 (1998-10-09)
Irganox 1330 (1,3,5-trimethyl-2,4,5-tris(3',5'-ditert-butyl)-4'-hydroxybenzyl)- benzene) (I), an alkylphenol compound used as antioxdiant in the production of high density polyethylene (HDPE), was the subject of this study. The quantity of I given up by polyethylene samples of different thickness to aqueous and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service