Skip to Content
Merck
All Photos(2)

Documents

407151

Sigma-Aldrich

2,6-Bis[(4S)-(−)-isopropyl-2-oxazolin-2-yl]pyridine

99%

Synonym(s):

(S,S)-2,2′-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline), (S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N3O2
CAS Number:
Molecular Weight:
301.38
Beilstein:
5824976
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]25/D −118°, c = 0.7 in methylene chloride

mp

155-157 °C (lit.)

SMILES string

CC(C)[C@H]1COC(=N1)c2cccc(n2)C3=N[C@H](CO3)C(C)C

InChI

1S/C17H23N3O2/c1-10(2)14-8-21-16(19-14)12-6-5-7-13(18-12)17-20-15(9-22-17)11(3)4/h5-7,10-11,14-15H,8-9H2,1-4H3/t14-,15-/m1/s1

InChI key

CSGQGLBCAHGJDR-HUUCEWRRSA-N

Application

Used as a ligand to prepare novel ruthenium-pyridine-carboxylate complexes as new catalytic systems for alkene epoxidation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nishiyama, H. et al.
Chemical Communications (Cambridge, England), 1863-1863 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service