Skip to Content
Merck
All Photos(2)

Key Documents

406961

Sigma-Aldrich

(+)-2,2′-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline]

97%

Synonym(s):

(R,R)-2,2′-Bis(4-phenyl-2-oxazolin-2-yl)propane, (R,R)-2,2′-Isopropylidenebis(4-phenyl-2-oxazoline)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N2O2
CAS Number:
Molecular Weight:
334.41
Beilstein:
4266906
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D +160°, c = 1 in ethanol

mp

56-58 °C (lit.)

functional group

ether
phenyl

storage temp.

−20°C

SMILES string

CC(C)(C1=N[C@@H](CO1)c2ccccc2)C3=N[C@@H](CO3)c4ccccc4

InChI

1S/C21H22N2O2/c1-21(2,19-22-17(13-24-19)15-9-5-3-6-10-15)20-23-18(14-25-20)16-11-7-4-8-12-16/h3-12,17-18H,13-14H2,1-2H3/t17-,18-/m0/s1

InChI key

JTNVCJCSECAMLD-ROUUACIJSA-N

Application

(+)-2,2′-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline] in the presence of copper iodide, can catalyze the asymmetric cyclopropanation reaction of phenyliodonium ylides with alkenes to form cyclopropane α-amino acid esters.
C2 symmetric ligand for enantioselective catalysis. Easily forms bidentate coordination complexes due to the strong affinity of the oxazoline nitrogen for various metals.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benoît Moreau et al.
Journal of the American Chemical Society, 127(51), 18014-18015 (2005-12-22)
A highly enantioselective (up to 97.5% ee) and diastereoselective (95:5 dr trans/cis) Cu(I)-catalyzed cyclopropanation of alkenes using phenyliodonium ylide generated in situ from iodosobenzene and methyl nitroacetate is reported. The cyclopropanation took place with high enantioselectivity for a wide range
Pfaltz, A.
Accounts of Chemical Research, 26, 339-339 (1993)
Bolm, C.
Angewandte Chemie (International Edition in English), 30, 542-542 (1991)
Gant, T.G. Meyers, A.I.
Tetrahedron, 50, 2297-2297 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service